missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,3,5-Trimethoxy-2-nitrobenzene, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1047322
Description
2,4,6-Trimethoxynitrobenzene is used as a chemicals, pharmaceutical intermediate, chemical research, application intermediate, pharmaceutical reagents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,4,6-Trimethoxynitrobenzene is used as a chemicals, pharmaceutical intermediate, chemical research, application intermediate, pharmaceutical reagents.
Solubility
Soluble in water.
Notes
Desiccate at room temperature. Incompatabile to strong oxidizing agents, strong acids and bases. Store in a cool, dry, well-ventilated place.
Specifications
1,3,5-Trimethoxy-2-nitrobenzene | |
150°C to 153°C | |
MFCD00016992 | |
2277262 | |
Soluble in water. | |
COC1=CC(=C(C(=C1)OC)[N+](=O)[O-])OC | |
213.189 | |
213.19 |
14227-18-0 | |
C9H11NO5 | |
100 g | |
2,4,6-trimethoxynitrobenzene, 1-nitro-2,4,6-trimethoxybenzene, benzene,1,3,5-trimethoxy-2-nitro, benzene, 1,3,5-trimethoxy-2-nitro, acmc-20ap5l, maybridge1_001624, 1,3,5-trimethoxy-2-nitro-benzene, # | |
VWYAWLZEMLQGJH-UHFFFAOYSA-N | |
1,3,5-trimethoxy-2-nitrobenzene | |
518903 | |
98% |
Chemical Identifiers
14227-18-0 | |
213.189 | |
VWYAWLZEMLQGJH-UHFFFAOYSA-N | |
1,3,5-trimethoxy-2-nitrobenzene |
C9H11NO5 | |
MFCD00016992 | |
518903 | |
COC1=CC(=C(C(=C1)OC)[N+](=O)[O-])OC |
Safety and Handling
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only