missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Trimethyl-1H-pyrazole-4-carboxylic acid, 97%, Thermo Scientific™

$275.69 - $275.69

Chemical Identifiers

CAS 1125-29-7
Molecular Formula C7H10N2O2
Molecular Weight (g/mol) 154.17
MDL Number MFCD00159648
InChI Key NOIOGQJFLIPRBI-UHFFFAOYSA-N
Synonym 1,3,5-trimethyl-1h-pyrazole-4-carboxylic acid, trimethyl-1h-pyrazole-4-carboxylic acid, trimethylpyrazole-4-carboxylic acid, 1,3,5-trimethyl-4-pyrazolecarboxylic acid, 1h-pyrazole-4-carboxylic acid, 1,3,5-trimethyl, akos pao-0337, rarechem al be 0403, art-chem-bb b006363, cambridge id 5264152, 1,3,5-trimethyl-1h-pyrazol-4-carboxylic acid
PubChem CID 736514
IUPAC Name 1,3,5-trimethylpyrazole-4-carboxylic acid
SMILES CN1N=C(C)C(C(O)=O)=C1C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC14701DA
SDS View Documents
Thermo Scientific Chemicals
CC14701DA
1 g Amber Glass Bottle
Each for $275.69
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1125-29-7
154.17
NOIOGQJFLIPRBI-UHFFFAOYSA-N
736514
CN1N=C(C)C(C(O)=O)=C1C
C7H10N2O2
MFCD00159648
1,3,5-trimethyl-1h-pyrazole-4-carboxylic acid, trimethyl-1h-pyrazole-4-carboxylic acid, trimethylpyrazole-4-carboxylic acid, 1,3,5-trimethyl-4-pyrazolecarboxylic acid, 1h-pyrazole-4-carboxylic acid, 1,3,5-trimethyl, akos pao-0337, rarechem al be 0403, art-chem-bb b006363, cambridge id 5264152, 1,3,5-trimethyl-1h-pyrazol-4-carboxylic acid
1,3,5-trimethylpyrazole-4-carboxylic acid

Specifications

1125-29-7
Amber Glass Bottle
MFCD00159648
1,3,5-trimethyl-1h-pyrazole-4-carboxylic acid, trimethyl-1h-pyrazole-4-carboxylic acid, trimethylpyrazole-4-carboxylic acid, 1,3,5-trimethyl-4-pyrazolecarboxylic acid, 1h-pyrazole-4-carboxylic acid, 1,3,5-trimethyl, akos pao-0337, rarechem al be 0403, art-chem-bb b006363, cambridge id 5264152, 1,3,5-trimethyl-1h-pyrazol-4-carboxylic acid
CN1N=C(C)C(C(O)=O)=C1C
154.17
154.17
1, 3, 5-Trimethyl-1H-pyrazole-4-carboxylic acid
97%
C7H10N2O2
1 g
NOIOGQJFLIPRBI-UHFFFAOYSA-N
1,3,5-trimethylpyrazole-4-carboxylic acid
736514
97%
Videos
Safety and Handling

Safety and Handling

Warning

SDS
missing translation for 'documents'

missing translation for 'documents'