missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Tris(trifluoromethyl)benzene 98.0+%, TCI America™

$497.70 - $497.70

Chemical Identifiers

CAS 729-81-7
Molecular Formula C9H3F9
Molecular Weight (g/mol) 282.109
MDL Number MFCD00054722
InChI Key ZMAUHKSOLPYPDB-UHFFFAOYSA-N
PubChem CID 123085
IUPAC Name 1,3,5-tris(trifluoromethyl)benzene
SMILES C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C(F)(F)F
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T28305G
View Documents
TCI America
T28305G
5 g
Each for $497.70
Only null left
 
Description

Description

Specifications

Chemical Identifiers

729-81-7
282.109
ZMAUHKSOLPYPDB-UHFFFAOYSA-N
1,3,5-tris(trifluoromethyl)benzene
C9H3F9
MFCD00054722
123085
C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C(F)(F)F

Specifications

729-81-7
120°C
MFCD00054722
1993
C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C(F)(F)F
282.109
282.11
Liquid
Colorless
C9H3F9
5 g
ZMAUHKSOLPYPDB-UHFFFAOYSA-N
1,3,5-tris(trifluoromethyl)benzene
123085
≥98.0% (GC)
1,3,5-Tris(trifluoromethyl)benzene
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'