missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,3-Benzenedisulfonyl Chloride 98.0+%, TCI America™
Supplier: TCI America B084825G
Specifications
1,3-Benzenedisulfonyl Chloride | |
62°C | |
C6H4Cl2O4S2 | |
25 g | |
1,3-benzenedisulfonyl chloride, benzene-1,3-disulfonyl dichloride, 1,3-benzenedisulfonyl dichloride, m-benzenedisulfonyl chloride, m-benzenedisulfonyl dichloride, m-phenylenedisulfonyl chloride, 1,3-benzenedisulfonyldichloride, 3-chlorosulfonylbenzenesulfonyl chloride, benzene-1,3-di sulphonyl chloride, acmc-209m5a | |
ClS(=O)(=O)C1=CC(=CC=C1)S(Cl)(=O)=O | |
275.11 | |
275.11 | |
Crystalline Powder |
585-47-7 | |
145°C | |
MFCD00041879 | |
3261 | |
ALIQZUMMPOYCIS-UHFFFAOYSA-N | |
benzene-1,3-disulfonyl dichloride | |
96240 | |
≥98.0% (HPLC,T) |
Chemical Identifiers
585-47-7 | |
275.11 | |
ALIQZUMMPOYCIS-UHFFFAOYSA-N | |
96240 | |
ClS(=O)(=O)C1=CC(=CC=C1)S(Cl)(=O)=O |
C6H4Cl2O4S2 | |
MFCD00041879 | |
1,3-benzenedisulfonyl chloride, benzene-1,3-disulfonyl dichloride, 1,3-benzenedisulfonyl dichloride, m-benzenedisulfonyl chloride, m-benzenedisulfonyl dichloride, m-phenylenedisulfonyl chloride, 1,3-benzenedisulfonyldichloride, 3-chlorosulfonylbenzenesulfonyl chloride, benzene-1,3-di sulphonyl chloride, acmc-209m5a | |
benzene-1,3-disulfonyl dichloride |
Safety and Handling
EINECSNumber : (3)-1899
TSCA : Yes
Recommended Storage : Refrigerator