missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(3-hydroxy-3-pentyl)benzene 90.0+%, TCI America™

$305.70 - $987.95

Chemical Identifiers

CAS 676465-94-4
Molecular Formula C16H26O2
Molecular Weight (g/mol) 250.382
InChI Key DBVBDLSFXMYKIU-UHFFFAOYSA-N
PubChem CID 44630004
IUPAC Name 3-[3-(3-hydroxypentan-3-yl)phenyl]pentan-3-ol
SMILES CCC(CC)(C1=CC(=CC=C1)C(CC)(CC)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B34365G
View Documents
TCI America
B34365G
5 g
Each for $305.70
Only null left
 
B343625G
View Documents
TCI America
B343625G
25 g
Each for $987.95
Only null left
 
Description

Description

Specifications

Chemical Identifiers

676465-94-4
250.382
44630004
CCC(CC)(C1=CC(=CC=C1)C(CC)(CC)O)O
C16H26O2
DBVBDLSFXMYKIU-UHFFFAOYSA-N
3-[3-(3-hydroxypentan-3-yl)phenyl]pentan-3-ol

Specifications

676465-94-4
C16H26O2
1,3-Bis(3-hydroxy-3-amyl)benzene, 1,3-Bis(1-ethyl-1-hydroxypropyl)benzene, 3,3′C-(m-Phenylene)di-3-pentanol, alpha,alpha,alpha′C,alpha′C-Tetraethyl-1,3-benzenedimethanol
CCC(CC)(C1=CC(=CC=C1)C(CC)(CC)O)O
250.382
250.38
Crystalline Powder
White-Yellow
5 g
DBVBDLSFXMYKIU-UHFFFAOYSA-N
3-[3-(3-hydroxypentan-3-yl)phenyl]pentan-3-ol
44630004
≥90.0% (GC)
1,3-Bis(3-hydroxy-3-pentyl)benzene
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'