missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,3-Bis(chloromethyl)tetramethyldisilazane 95.0+%, TCI America™
Supplier: TCI America B09905G
Specifications
1,3-Bis(chloromethyl)tetramethyldisilazane | |
Yellow | |
C6H17Cl2NSi2 | |
5 g | |
Tetramethyl-1,3-bis(chloromethyl)disilazane | |
C[Si](C)(CCl)N[Si](C)(C)CCl | |
230.279 | |
230.28 | |
Liquid |
14579-91-0 | |
72°C | |
MFCD00000876 | |
1760 | |
SXSNZRHGAMVNJE-UHFFFAOYSA-N | |
chloro-[[[chloromethyl(dimethyl)silyl]amino]-dimethylsilyl]methane | |
26740 | |
≥95.0% (GC) |
Chemical Identifiers
14579-91-0 | |
230.279 | |
SXSNZRHGAMVNJE-UHFFFAOYSA-N | |
26740 | |
C[Si](C)(CCl)N[Si](C)(C)CCl |
C6H17Cl2NSi2 | |
MFCD00000876 | |
Tetramethyl-1,3-bis(chloromethyl)disilazane | |
chloro-[[[chloromethyl(dimethyl)silyl]amino]-dimethylsilyl]methane |
Safety and Handling
RTECSNumber : JM9210000
TSCA : Yes