missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(methylthio)-2-methoxypropane 97.0+%, TCI America™

$398.90 - $398.90

Chemical Identifiers

CAS 31805-84-2
Molecular Formula C6H14OS2
Molecular Weight (g/mol) 166.297
MDL Number MFCD00026040
InChI Key MOISORXUYSHXRU-UHFFFAOYSA-N
Synonym 1,3-bis methylthio-2-methoxypropane, 2-methoxy-1,3-bis methylsulfanyl propane, 2-methoxy-1,3-bis methylthio propane, 1,3-bis-methylthio-2-methoxypropane, propane, 2-methoxy-1,3-bis methylthio, acmc-1cktt, moisorxuyshxru-uhfffaoysa, propane,2-methoxy-1,3-bis methylthio, 2-methoxy-1,3-bis methylsulfanyl-propane, 2-methoxy-1,3-bis methylsulfanyl propane #
PubChem CID 520544
IUPAC Name 2-methoxy-1,3-bis(methylsulfanyl)propane
SMILES COC(CSC)CSC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B17095G
View Documents
TCI America
B17095G
5 g
Each for $398.90
Only null left
 
Description

Description

Specifications

Chemical Identifiers

31805-84-2
166.297
MOISORXUYSHXRU-UHFFFAOYSA-N
520544
COC(CSC)CSC
C6H14OS2
MFCD00026040
1,3-bis methylthio-2-methoxypropane, 2-methoxy-1,3-bis methylsulfanyl propane, 2-methoxy-1,3-bis methylthio propane, 1,3-bis-methylthio-2-methoxypropane, propane, 2-methoxy-1,3-bis methylthio, acmc-1cktt, moisorxuyshxru-uhfffaoysa, propane,2-methoxy-1,3-bis methylthio, 2-methoxy-1,3-bis methylsulfanyl-propane, 2-methoxy-1,3-bis methylsulfanyl propane #
2-methoxy-1,3-bis(methylsulfanyl)propane

Specifications

31805-84-2
97°C
MFCD00026040
1,3-bis methylthio-2-methoxypropane, 2-methoxy-1,3-bis methylsulfanyl propane, 2-methoxy-1,3-bis methylthio propane, 1,3-bis-methylthio-2-methoxypropane, propane, 2-methoxy-1,3-bis methylthio, acmc-1cktt, moisorxuyshxru-uhfffaoysa, propane,2-methoxy-1,3-bis methylthio, 2-methoxy-1,3-bis methylsulfanyl-propane, 2-methoxy-1,3-bis methylsulfanyl propane #
COC(CSC)CSC
166.297
166.30
Liquid
Yellow
C6H14OS2
5 g
MOISORXUYSHXRU-UHFFFAOYSA-N
2-methoxy-1,3-bis(methylsulfanyl)propane
520544
≥97.0% (GC)
1,3-Bis(methylthio)-2-methoxypropane
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'