missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3-Butadiene (ca. 13% in Tetrahydrofuran, ca. 2mol/L), TCI America™

Supplier:  TCI America B4835100ML

Catalog No. B4835100ML


Explore available promotions

Description

Description

Specifications

Specifications

1,3-Butadiene (ca. 13% in Tetrahydrofuran, ca. 2mol/L)
Yellow
MFCD00008659
1993
KAKZBPTYRLMSJV-UHFFFAOYSA-N
buta-1,3-diene
7845
54.09
106-99-0
C4H6
100 mL
1,3-butadiene, divinyl, butadiene, vinylethylene, biethylene, erythrene, bivinyl, pyrrolylene, butadieen, butadien
C=CC=C
54.092
CHEBI:39478
Liquid

Chemical Identifiers

106-99-0
54.092
KAKZBPTYRLMSJV-UHFFFAOYSA-N
7845
buta-1,3-diene
C4H6
MFCD00008659
1,3-butadiene, divinyl, butadiene, vinylethylene, biethylene, erythrene, bivinyl, pyrrolylene, butadieen, butadien
CHEBI:39478
C=CC=C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : EI9275000

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions