missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,3-Butadiene Monoepoxide 97.0+%, TCI America™
Supplier: TCI America E03375G
Specifications
1,3-Butadiene Monoepoxide | |
-135°C | |
67°C | |
MFCD00005149 | |
1993 | |
GXBYFVGCMPJVJX-UHFFFAOYSA-N | |
2-ethenyloxirane | |
13586 | |
≥97.0% (GC) |
930-22-3 | |
Yellow | |
C4H6O | |
5 g | |
butadiene monoxide, oxirane, ethenyl, 3,4-epoxy-1-butene, 2-vinyloxirane, butadiene epoxide, 1,2-epoxy-3-butene, vinyloxirane, butadiene oxide, butadiene monoepoxide, butadiene monooxide | |
C=CC1CO1 | |
70.091 | |
70.09 | |
Liquid |
Chemical Identifiers
930-22-3 | |
70.091 | |
GXBYFVGCMPJVJX-UHFFFAOYSA-N | |
13586 | |
C=CC1CO1 |
C4H6O | |
MFCD00005149 | |
butadiene monoxide, oxirane, ethenyl, 3,4-epoxy-1-butene, 2-vinyloxirane, butadiene epoxide, 1,2-epoxy-3-butene, vinyloxirane, butadiene oxide, butadiene monoepoxide, butadiene monooxide | |
2-ethenyloxirane |
Safety and Handling
RTECSNumber : EM7350000
TSCA : Yes