missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(±)-1,3-Butanediol, 99%, Extra Pure, Thermo Scientific Chemicals
$66.94 - $605.68
Chemical Identifiers
CAS | 107-88-0 |
---|---|
Molecular Formula | C4H10O2 |
Molecular Weight (g/mol) | 90.12 |
MDL Number | MFCD00004554 |
InChI Key | PUPZLCDOIYMWBV-UHFFFAOYSA-N |
Synonym | 1,3-butanediol, 1,3-butylene glycol, 1,3-dihydroxybutane, methyltrimethylene glycol, 1,3-butandiol, 1-methyl-1,3-propanediol, beta-butylene glycol, 1,3 butylene glycol, 1,3-butylenglykol, rs-1,3-butandiol |
PubChem CID | 7896 |
ChEBI | CHEBI:52683 |
IUPAC Name | butane-1,3-diol |
SMILES | CC(CCO)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC107622500
|
Thermo Scientific Chemicals
107622500 |
250 mL | Glass bottle |
Each for $66.94
|
|
|||||
AC107620010
|
Thermo Scientific Chemicals
107620010 |
1 L | Glass bottle |
Each for $147.78
Case of 4 Each for $0.00
|
|
|||||
AC107620025
|
Thermo Scientific Chemicals
107620025 |
2.5 L | Glass bottle |
Each for $325.44
|
|
|||||
AC107620100
|
Thermo Scientific Chemicals
107620100 |
10 L | Plastic drum |
Each for $605.68
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
107-88-0 | |
90.12 | |
PUPZLCDOIYMWBV-UHFFFAOYSA-N | |
7896 | |
butane-1,3-diol |
C4H10O2 | |
MFCD00004554 | |
1,3-butanediol, 1,3-butylene glycol, 1,3-dihydroxybutane, methyltrimethylene glycol, 1,3-butandiol, 1-methyl-1,3-propanediol, beta-butylene glycol, 1,3 butylene glycol, 1,3-butylenglykol, rs-1,3-butandiol | |
CHEBI:52683 | |
CC(CCO)O |
Specifications
107-88-0 | |
100.0 | |
Brown to Colorless | |
203.0°C to 204.0°C | |
0.05% max. | |
98.5% min. (GC) | |
C4H10O2 | |
CH3CH(OH)CH2CH2OH | |
250 mL | |
1.01 | |
1,3-butanediol, 1,3-butylene glycol, 1,3-dihydroxybutane, methyltrimethylene glycol, 1,3-butandiol, 1-methyl-1,3-propanediol, beta-butylene glycol, 1,3 butylene glycol, 1,3-butylenglykol, rs-1,3-butandiol | |
PUPZLCDOIYMWBV-UHFFFAOYSA-N | |
butane-1,3-diol | |
7896 | |
CHEBI:52683 | |
99% | |
Liquid |
98.5 | |
-54.0°C | |
1.0100g/mL | |
109°C | |
Authentic | |
Glass bottle | |
1.4385 to 1.4405 | |
MFCD00004554 | |
01, 477 | |
15, 1570 | |
Solubility in water: soluble. Other solubilities: soluble in acetone, methyl ethyl ketone, ethanol, slightly soluble in ether, practically insoluble in aliphatic hydrocarbons,, benzene, toluene and ccl4. | |
CC(CCO)O | |
90.12 | |
24.6 mm2/s (50°C) | |
90.12 | |
Extra Pure | |
(±)-1,3-Butanediol |
Safety and Handling
EINECSNumber : 203-529-7
RUO – Research Use Only