missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3-Cyclohexanediol, cis + trans, 98%, Thermo Scientific Chemicals

$64.29 - $231.92

Chemical Identifiers

CAS 504-01-8
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00039458
InChI Key RLMGYIOTPQVQJR-UHFFFAOYSA-N
Synonym 1,3-cyclohexanediol, resorcitol, 1,3-dihydroxycyclohexane, 1,3-benzenediol, hexahydro, cis-1,3-cyclohexandiol, trans-1,3-cyclohexanediol, trans-1,3-cyclohexanol, 1,3-cyclohexanediol,c&t, acmc-209ll9, acmc-209po7
PubChem CID 10433
IUPAC Name cyclohexane-1,3-diol
SMILES C1CC(CC(C1)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0411006
View Documents
Thermo Scientific Chemicals
L0411006
5 g
Each for $64.29
 
AAL0411014
View Documents
Thermo Scientific Chemicals
L0411014
25 g
Each for $231.92
 
Description

Description

1,3-Cyclohexanediol is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Cyclohexanediol is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

504-01-8
116.16
RLMGYIOTPQVQJR-UHFFFAOYSA-N
10433
C1CC(CC(C1)O)O
C6H12O2
MFCD00039458
1,3-cyclohexanediol, resorcitol, 1,3-dihydroxycyclohexane, 1,3-benzenediol, hexahydro, cis-1,3-cyclohexandiol, trans-1,3-cyclohexanediol, trans-1,3-cyclohexanol, 1,3-cyclohexanediol,c&t, acmc-209ll9, acmc-209po7
cyclohexane-1,3-diol

Specifications

504-01-8
246°C to 247°C
MFCD00039458
2036654
Soluble in water.
C1CC(CC(C1)O)O
116.16
116.16
1,3-Cyclohexanediol, Cis + Trans
∼30°C
C6H12O2
5 g
1,3-cyclohexanediol, resorcitol, 1,3-dihydroxycyclohexane, 1,3-benzenediol, hexahydro, cis-1,3-cyclohexandiol, trans-1,3-cyclohexanediol, trans-1,3-cyclohexanol, 1,3-cyclohexanediol,c&t, acmc-209ll9, acmc-209po7
RLMGYIOTPQVQJR-UHFFFAOYSA-N
cyclohexane-1,3-diol
10433
98%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 207-979-5

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only