missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3-Cyclopentanediol, 95%, mixture of cis and trans, Thermo Scientific Chemicals

$188.56 - $188.56

Chemical Identifiers

CAS 59719-74-3
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00001367
InChI Key NUUPJBRGQCEZSI-UHFFFAOYSA-N
Synonym 1,3-cyclopentanediol, 1,3-cyclopentanediol, cis, 1,3-cyclopentanediol, mixture of cis and trans, 1,3-cyclopentandiol, 1,4-cyclopentanediol, cyclopentan-1,3-diol, 3-hydroxy cylcopentanol, cyclopentane 1,3-diol, acmc-20ap4n, 1,3-dihydroxycyclopentane
PubChem CID 100165
IUPAC Name cyclopentane-1,3-diol
SMILES C1CC(CC1O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Qty  
Catalog Number Mfr. No. Quantity Packaging Price Qty  
AC185490010
SDS missing translation for 'viewDocuments'
Thermo Scientific Chemicals
185490010
1 g Glass bottle
Each for $188.56
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

59719-74-3
102.13
NUUPJBRGQCEZSI-UHFFFAOYSA-N
100165
C1CC(CC1O)O
C5H10O2
MFCD00001367
1,3-cyclopentanediol, 1,3-cyclopentanediol, cis, 1,3-cyclopentanediol, mixture of cis and trans, 1,3-cyclopentandiol, 1,4-cyclopentanediol, cyclopentan-1,3-diol, 3-hydroxy cylcopentanol, cyclopentane 1,3-diol, acmc-20ap4n, 1,3-dihydroxycyclopentane
cyclopentane-1,3-diol

Specifications

59719-74-3
100.0
Colorless to Yellow
80.0°C to 85.0°C (0.1 mmHg)
Authentic
Glass bottle
1.483
MFCD00001367
06, III, 4057
1,3-cyclopentanediol, 1,3-cyclopentanediol, cis, 1,3-cyclopentanediol, mixture of cis and trans, 1,3-cyclopentandiol, 1,4-cyclopentanediol, cyclopentan-1,3-diol, 3-hydroxy cylcopentanol, cyclopentane 1,3-diol, acmc-20ap4n, 1,3-dihydroxycyclopentane
NUUPJBRGQCEZSI-UHFFFAOYSA-N
cyclopentane-1,3-diol
100165
95%
1,3-Cyclopentanediol, Mixture of cis and trans
94.0
40.0°C
1.0900g/mL
>110°C
94% min. (GC)
C5H10O2
C5H8(OH)2
1 g
1.09
Solubility in water: partially soluble.
C1CC(CC1O)O
102.13
102.13
Liquid After Melting
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

GHS Signal Word: Warning

EINECSNumber : 261-880-1

SDS
Documents

Documents

RUO – Research Use Only