missing translation for 'onlineSavingsMsg'
Learn More

1,3-Di-BOC-2-(trifluoromethylsulfonyl)guanidine, 96%

CAS: 207857-15-6 | C12H20F3N3O6S | 391.36 g/mol

$1108.26 - $1108.26

Chemical Identifiers

CAS 207857-15-6
Molecular Formula C12H20F3N3O6S
Molecular Weight (g/mol) 391.36
InChI Key GOQZIPJCBUYLIR-UHFFFAOYSA-N
Synonym 1,3-di-boc-2-trifluoromethylsulfonyl guanidine, goodman's reagent, n,n'-di-boc-n-triflylguanidine, 1,3-bis tert-butoxycarbonyl-2-trifluoromethanesulfonyl guanidine, 1,3-bis tert-butoxycarbonyl-2-trifluoromethylsulfonyl guanidine, 1,3-di-boc-2-trifluoromethanesulfonyl guanidine, tert-butyl n-n-2-methylpropan-2-yl oxycarbonyl-n'-trifluoromethylsulfonyl carbamimidoyl carbamate, tert-butyl n-tert-butoxycarbonyl amino trifluoromethanesulfonylimino methyl carbamate, tert-butyl n-tert-butoxycarbonyl-n'-trifluoromethyl sulphonyl carbamimidoyl carbamate
PubChem CID 3694570
IUPAC Name tert-butyl N-[N-[(2-methylpropan-2-yl)oxycarbonyl]-N'-(trifluoromethylsulfonyl)carbamimidoyl]carbamate
SMILES CC(C)(C)OC(=O)NC(=NS(=O)(=O)C(F)(F)F)NC(=O)OC(C)(C)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC399750050
SDS View Documents
Thermo Scientific Chemicals
399750050
5 g Glass Bottle
Each for $1,108.26
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

207857-15-6
391.36
1,3-di-boc-2-trifluoromethylsulfonyl guanidine, goodman's reagent, n,n'-di-boc-n-triflylguanidine, 1,3-bis tert-butoxycarbonyl-2-trifluoromethanesulfonyl guanidine, 1,3-bis tert-butoxycarbonyl-2-trifluoromethylsulfonyl guanidine, 1,3-di-boc-2-trifluoromethanesulfonyl guanidine, tert-butyl n-n-2-methylpropan-2-yl oxycarbonyl-n'-trifluoromethylsulfonyl carbamimidoyl carbamate, tert-butyl n-tert-butoxycarbonyl amino trifluoromethanesulfonylimino methyl carbamate, tert-butyl n-tert-butoxycarbonyl-n'-trifluoromethyl sulphonyl carbamimidoyl carbamate
tert-butyl N-[N-[(2-methylpropan-2-yl)oxycarbonyl]-N'-(trifluoromethylsulfonyl)carbamimidoyl]carbamate
C12H20F3N3O6S
GOQZIPJCBUYLIR-UHFFFAOYSA-N
3694570
CC(C)(C)OC(=O)NC(=NS(=O)(=O)C(F)(F)F)NC(=O)OC(C)(C)C

Specifications

111°C to 113°C
97.5% min. (HPLC)
Glass Bottle
GOQZIPJCBUYLIR-UHFFFAOYSA-N
tert-butyl N-[N-[(2-methylpropan-2-yl)oxycarbonyl]-N'-(trifluoromethylsulfonyl)carbamimidoyl]carbamate
391.36
391.36
1, 3-Di-BOC-2-(trifluoromethylsulfonyl)guanidine
207857-15-6
C12H20F3N3O6S
1,3-di-boc-2-trifluoromethylsulfonyl guanidine, goodman's reagent, n,n'-di-boc-n-triflylguanidine, 1,3-bis tert-butoxycarbonyl-2-trifluoromethanesulfonyl guanidine, 1,3-bis tert-butoxycarbonyl-2-trifluoromethylsulfonyl guanidine, 1,3-di-boc-2-trifluoromethanesulfonyl guanidine, tert-butyl n-n-2-methylpropan-2-yl oxycarbonyl-n'-trifluoromethylsulfonyl carbamimidoyl carbamate, tert-butyl n-tert-butoxycarbonyl amino trifluoromethanesulfonylimino methyl carbamate, tert-butyl n-tert-butoxycarbonyl-n'-trifluoromethyl sulphonyl carbamimidoyl carbamate
CC(C)(C)OC(=O)NC(=NS(=O)(=O)C(F)(F)F)NC(=O)OC(C)(C)C
5 g
3694570
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only