missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Diacetoxyacetone 98.0+%, TCI America™
Supplier: TCI America D284325G
Specifications
| 1,3-Diacetoxyacetone | |
| 50°C | |
| MFCD01310655 | |
| 1,3-Diacetoxy-2-propanone | |
| CC(=O)OCC(=O)COC(=O)C | |
| 174.15 | |
| 174.15 | |
| Crystalline Powder |
| 6946-10-7 | |
| C7H10O5 | |
| 25 g | |
| PZVCVSQSQHGBNE-UHFFFAOYSA-N | |
| (3-acetyloxy-2-oxopropyl) acetate | |
| 243418 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 6946-10-7 | |
| 174.15 | |
| PZVCVSQSQHGBNE-UHFFFAOYSA-N | |
| 243418 | |
| CC(=O)OCC(=O)COC(=O)C |
| C7H10O5 | |
| MFCD01310655 | |
| 1,3-Diacetoxy-2-propanone | |
| (3-acetyloxy-2-oxopropyl) acetate |
Safety and Handling
TSCA : No