Learn More
1,3-Diamino-2-propanol, 97%
CAS: 616-29-5 | C3H10N2O | 90.126 g/mol
$129.92 - $428.97
Chemical Identifiers
| CAS | 616-29-5 |
|---|---|
| Molecular Formula | C3H10N2O |
| Molecular Weight (g/mol) | 90.126 |
| MDL Number | MFCD00008142 |
| InChI Key | UYBWIEGTWASWSR-UHFFFAOYSA-N |
| Synonym | 1,3-diamino-2-propanol, 1,3-diamino-2-hydroxypropane, diaminoisopropanol, 2-hydroxy-1,3-diaminopropane, 2-propanol, 1,3-diamino, 2-hydroxy-1,3-propanediamine, 1,3-diamino-2-hydroxy-propane, 2-propanol,3-diamino, acmc-1b1xv, 1,3 diamino-propan-2-ol |
| PubChem CID | 61157 |
| IUPAC Name | 1,3-diaminopropan-2-ol |
| SMILES | C(C(CN)O)N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1961809
|
Thermo Scientific Chemicals
A1961809 |
10 g |
Each for $129.92
|
|
|||||
|
AAA1961818
|
Thermo Scientific Chemicals
A1961818 |
50 g |
Each for $428.97
|
|
|||||
Description
It finds its application as the formaldehyde absorbent, carbon dioxide and acid gas absorption. It is also used as a pharmaceutical intermediate and in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt finds its application as the formaldehyde absorbent, carbon dioxide and acid gas absorption. It is also used as a pharmaceutical intermediate and in organic synthesis.
Solubility
Very soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
| 616-29-5 | |
| 90.126 | |
| UYBWIEGTWASWSR-UHFFFAOYSA-N | |
| 61157 | |
| C(C(CN)O)N |
| C3H10N2O | |
| MFCD00008142 | |
| 1,3-diamino-2-propanol, 1,3-diamino-2-hydroxypropane, diaminoisopropanol, 2-hydroxy-1,3-diaminopropane, 2-propanol, 1,3-diamino, 2-hydroxy-1,3-propanediamine, 1,3-diamino-2-hydroxy-propane, 2-propanol,3-diamino, acmc-1b1xv, 1,3 diamino-propan-2-ol | |
| 1,3-diaminopropan-2-ol |
Specifications
| 616-29-5 | |
| 235°C | |
| C3H10N2O | |
| 10 g | |
| 1,3-diamino-2-propanol, 1,3-diamino-2-hydroxypropane, diaminoisopropanol, 2-hydroxy-1,3-diaminopropane, 2-propanol, 1,3-diamino, 2-hydroxy-1,3-propanediamine, 1,3-diamino-2-hydroxy-propane, 2-propanol,3-diamino, acmc-1b1xv, 1,3 diamino-propan-2-ol | |
| UYBWIEGTWASWSR-UHFFFAOYSA-N | |
| 1,3-diaminopropan-2-ol | |
| 61157 | |
| 97% |
| 40°C to 45°C | |
| >110°C (230°F) | |
| MFCD00008142 | |
| 741859 | |
| Very soluble in water. | |
| C(C(CN)O)N | |
| 90.126 | |
| 90.13 | |
| 1,3-Diamino-2-propanol |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 210-474-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only