Learn More
1,3-Dichloro-2-butene, cis + trans, 98%, stab. with BHT, Thermo Scientific Chemicals
$71.96 - $199.05
Chemical Identifiers
CAS | 926-57-8 |
---|---|
Molecular Formula | C4H6Cl2 |
Molecular Weight (g/mol) | 124.99 |
MDL Number | MFCD00000861 |
InChI Key | WLIADPFXSACYLS-DUXPYHPUSA-N |
Synonym | 1,3-dichlorobutylene, unii-pwg1e5mrlj, 1,3-dichloro-2-butene, trans-1,3-dichlorobutene-2, 1,3-dichlorobut-2-ene, pwg1e5mrlj, 2-butene, 1,3-dichloro-, e, 2-butene, 1,3-dichloro, e-1,3-dichloro-2-butene, 1,3-dichlorobutene-2 |
PubChem CID | 643911 |
SMILES | C\C(Cl)=C/CCl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL2033406
|
Thermo Scientific Chemicals
L2033406 |
5 g |
Each for $71.96
|
|
|||||
AAL2033414
|
Thermo Scientific Chemicals
L2033414 |
25 g |
Each for $199.05
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
926-57-8 | |
124.99 | |
WLIADPFXSACYLS-DUXPYHPUSA-N | |
643911 |
C4H6Cl2 | |
MFCD00000861 | |
1,3-dichlorobutylene, unii-pwg1e5mrlj, 1,3-dichloro-2-butene, trans-1,3-dichlorobutene-2, 1,3-dichlorobut-2-ene, pwg1e5mrlj, 2-butene, 1,3-dichloro-, e, 2-butene, 1,3-dichloro, e-1,3-dichloro-2-butene, 1,3-dichlorobutene-2 | |
C\C(Cl)=C/CCl |
Specifications
926-57-8 | |
125°C to 129°C | |
C4H6Cl2 | |
MFCD00000861 | |
UN1992 | |
Moisture sensitive | |
WLIADPFXSACYLS-DUXPYHPUSA-N | |
(2E)-1,3-dichlorobut-2-ene | |
643911 | |
98% |
1.161 | |
33°C (91°F) | |
1.473 | |
5 g | |
906754 | |
1,3-dichlorobutylene, unii-pwg1e5mrlj, 1,3-dichloro-2-butene, trans-1,3-dichlorobutene-2, 1,3-dichlorobut-2-ene, pwg1e5mrlj, 2-butene, 1,3-dichloro-, e, 2-butene, 1,3-dichloro, e-1,3-dichloro-2-butene, 1,3-dichlorobutene-2 | |
C\C(Cl)=C/CCl | |
124.99 | |
125 | |
1,3-Dichloro-2-butene, Cis + Trans |
Safety and Handling
GHS H Statement
H331-H226-H302-H315-H319-H335
Toxic if inhaled.
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H226-H301+H331-H312-H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 213-138-3
RTECSNumber : EM4760000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only