missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3-Dichloro-5,5-dimethylhydantoin 97.0+%, TCI America™

$43.44 - $120.48

Chemical Identifiers

CAS 118-52-5
Molecular Formula C5H6Cl2N2O2
Molecular Weight (g/mol) 197.015
MDL Number MFCD00003190
InChI Key KEQGZUUPPQEDPF-UHFFFAOYSA-N
Synonym 1,3-dichloro-5,5-dimethylhydantoin, dantoin, dactin, daktin, dichlorantin, omchlor, halane, hydan, hydantoin, dichlorodimethyl, hydan antiseptic
PubChem CID 8360
IUPAC Name 1,3-dichloro-5,5-dimethylimidazolidine-2,4-dione
SMILES CC1(C(=O)N(C(=O)N1Cl)Cl)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D178325G
View Documents
TCI America
D178325G
25 g
Each for $43.44
 
D1783100G
View Documents
TCI America
D1783100G
100 g
Each for $57.27
 
D1783500G
View Documents
TCI America
D1783500G
500 g
Each for $120.48
 
Description

Description

Specifications

Chemical Identifiers

118-52-5
197.015
KEQGZUUPPQEDPF-UHFFFAOYSA-N
8360
CC1(C(=O)N(C(=O)N1Cl)Cl)C
C5H6Cl2N2O2
MFCD00003190
1,3-dichloro-5,5-dimethylhydantoin, dantoin, dactin, daktin, dichlorantin, omchlor, halane, hydan, hydantoin, dichlorodimethyl, hydan antiseptic
1,3-dichloro-5,5-dimethylimidazolidine-2,4-dione

Specifications

118-52-5
White
MFCD00003190
3085
KEQGZUUPPQEDPF-UHFFFAOYSA-N
1,3-dichloro-5,5-dimethylimidazolidine-2,4-dione
8360
≥97.0% (T)
1,3-Dichloro-5,5-dimethylhydantoin
132°C
C5H6Cl2N2O2
25 g
1,3-dichloro-5,5-dimethylhydantoin, dantoin, dactin, daktin, dichlorantin, omchlor, halane, hydan, hydantoin, dichlorodimethyl, hydan antiseptic
CC1(C(=O)N(C(=O)N1Cl)Cl)C
197.015
197.02
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (9)-0580

RTECSNumber : MU0700000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'