Learn More
1,3-Dichlorobenzene, 98%, Thermo Scientific Chemicals
$99.96 - $288.14
Chemical Identifiers
CAS | 541-73-1 |
---|---|
Molecular Formula | C6H4Cl2 |
Molecular Weight (g/mol) | 146.998 |
MDL Number | MFCD00000573 |
InChI Key | ZPQOPVIELGIULI-UHFFFAOYSA-N |
Synonym | m-dichlorobenzene, m-phenylene dichloride, m-dichlorobenzol, meta-dichlorobenzene, benzene, 1,3-dichloro, metadichlorobenzene, benzene, m-dichloro, m-phenylenedichloride, m-dcb, 3-dichlorobenzene |
PubChem CID | 10943 |
ChEBI | CHEBI:36693 |
IUPAC Name | 1,3-dichlorobenzene |
SMILES | C1=CC(=CC(=C1)Cl)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1368830
|
Thermo Scientific Chemicals
A1368830 |
250 g |
Each for $99.96
|
|
|||||
AAA1368836
|
Thermo Scientific Chemicals
A1368836 |
500 g |
Each for $172.05
|
|
|||||
AAA136880B
|
Thermo Scientific Chemicals
A136880B |
1000 g |
Each for $288.14
|
|
|||||
Description
1,3-Dichlorobenzene is used in medicine, and dyes. And it is also used to study the effect of solvent vapor pressure on the vertical nanowire of C60 molecules1. 1,3-Dichlorobenzene, a chlorinated aromatic hydrocarbon, can be detected in environmental samples using an advanced Fourier transform infrared spectroscopy-attenuated total reflectance (FTIR-ATR) sensor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,3-Dichlorobenzene is used in medicine, and dyes. And it is also used to study the effect of solvent vapor pressure on the vertical nanowire of C60 molecules1. 1,3-Dichlorobenzene, a chlorinated aromatic hydrocarbon, can be detected in environmental samples using an advanced Fourier transform infrared spectroscopy-attenuated total reflectance (FTIR-ATR) sensor.
Solubility
Difficult to mix.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Chemical Identifiers
541-73-1 | |
146.998 | |
ZPQOPVIELGIULI-UHFFFAOYSA-N | |
10943 | |
1,3-dichlorobenzene |
C6H4Cl2 | |
MFCD00000573 | |
m-dichlorobenzene, m-phenylene dichloride, m-dichlorobenzol, meta-dichlorobenzene, benzene, 1,3-dichloro, metadichlorobenzene, benzene, m-dichloro, m-phenylenedichloride, m-dcb, 3-dichlorobenzene | |
CHEBI:36693 | |
C1=CC(=CC(=C1)Cl)Cl |
Specifications
541-73-1 | |
1.288 | |
63°C (145°F) | |
1.546 | |
250 g | |
956618 | |
m-dichlorobenzene, m-phenylene dichloride, m-dichlorobenzol, meta-dichlorobenzene, benzene, 1,3-dichloro, metadichlorobenzene, benzene, m-dichloro, m-phenylenedichloride, m-dcb, 3-dichlorobenzene | |
ZPQOPVIELGIULI-UHFFFAOYSA-N | |
1,3-dichlorobenzene | |
10943 | |
147 | |
1,3-Dichlorobenzene |
-25°C | |
173°C | |
C6H4Cl2 | |
MFCD00000573 | |
UN3082 | |
14,3055 | |
Difficult to mix. | |
C1=CC(=CC(=C1)Cl)Cl | |
146.998 | |
CHEBI:36693 | |
98% |
Safety and Handling
GHS H Statement
H302-H227
Harmful if swallowed.
Combustible liquid.
P210-P233-P240-P243-P261-P264b-P270-P271-P280-P301+P312-P304+P340-P312-P330-P374-P501c
H227-H302-H332
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber : 208-792-1
RTECSNumber : CZ4499000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only