Learn More
1,3-Dicyclohexylimidazolium tetrafluoroborate, 97%
CAS: 286014-38-8 | C15H25BF4N2 | 320.18 g/mol
$165.77 - $639.04
Chemical Identifiers
| CAS | 286014-38-8 |
|---|---|
| Molecular Formula | C15H25BF4N2 |
| Molecular Weight (g/mol) | 320.18 |
| MDL Number | MFCD04973310 |
| InChI Key | CQHXJIHJFMBBQA-UHFFFAOYSA-N |
| Synonym | 1,3-dicyclohexyl-1h-imidazol-3-ium tetrafluoroborate, 1,3-dicyclohexylimidazolium tetrafluoroborate, 1,3-dicyclohexyl-imidazolium tetrafluoroborate, 1,3-bis cyclohexyl imidazolium tetrafluoroborate, 1,3-dicyclohexyl-1??-imidazol-1-ylium tetrafluoroborate, acmc-1cq4s, 1,3-dicyclohexylimidazoliumtetrafluoroboratesalt, 1,3-dicyclohexylimidazolium tetrafluoroborate salt |
| PubChem CID | 16218169 |
| IUPAC Name | 1,3-dicyclohexyl-3H-1λâµ-imidazol-1-ylium; tetrafluoroboranuide |
| SMILES | F[B-](F)(F)F.C1CCC(CC1)N1C=C[N+](=C1)C1CCCCC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH3663303
|
Thermo Scientific Chemicals
H3663303 |
1 g |
Each for $165.77
|
|
|||||
|
AAH3663306
|
Thermo Scientific Chemicals
H3663306 |
5 g |
Each for $639.04
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 286014-38-8 | |
| 320.18 | |
| CQHXJIHJFMBBQA-UHFFFAOYSA-N | |
| 16218169 | |
| F[B-](F)(F)F.C1CCC(CC1)N1C=C[N+](=C1)C1CCCCC1 |
| C15H25BF4N2 | |
| MFCD04973310 | |
| 1,3-dicyclohexyl-1h-imidazol-3-ium tetrafluoroborate, 1,3-dicyclohexylimidazolium tetrafluoroborate, 1,3-dicyclohexyl-imidazolium tetrafluoroborate, 1,3-bis cyclohexyl imidazolium tetrafluoroborate, 1,3-dicyclohexyl-1??-imidazol-1-ylium tetrafluoroborate, acmc-1cq4s, 1,3-dicyclohexylimidazoliumtetrafluoroboratesalt, 1,3-dicyclohexylimidazolium tetrafluoroborate salt | |
| 1,3-dicyclohexyl-3H-1λâµ-imidazol-1-ylium; tetrafluoroboranuide |
Specifications
| 286014-38-8 | |
| Brown to White | |
| MFCD04973310 | |
| Moisture sensitive | |
| CQHXJIHJFMBBQA-UHFFFAOYSA-N | |
| 1,3-dicyclohexyl-3H-1λâµ-imidazol-1-ylium; tetrafluoroboranuide | |
| 16218169 | |
| 97% | |
| 1,3-Dicyclohexylimidazolium tetrafluoroborate |
| 173°C to 177°C | |
| C15H25BF4N2 | |
| 1 g | |
| 1,3-dicyclohexyl-1h-imidazol-3-ium tetrafluoroborate, 1,3-dicyclohexylimidazolium tetrafluoroborate, 1,3-dicyclohexyl-imidazolium tetrafluoroborate, 1,3-bis cyclohexyl imidazolium tetrafluoroborate, 1,3-dicyclohexyl-1??-imidazol-1-ylium tetrafluoroborate, acmc-1cq4s, 1,3-dicyclohexylimidazoliumtetrafluoroboratesalt, 1,3-dicyclohexylimidazolium tetrafluoroborate salt | |
| F[B-](F)(F)F.C1CCC(CC1)N1C=C[N+](=C1)C1CCCCC1 | |
| 320.18 | |
| 320.18 | |
| Powder |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
RTECSNumber : NONE
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only