missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3-Dimethyl-1H-pyrazole-5-carboxylic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 5744-56-9
Molecular Formula C6H8N2O2
Molecular Weight (g/mol) 140.14
MDL Number MFCD00159636
InChI Key QRWZFUBHOQWUGH-UHFFFAOYSA-N
Synonym 1,3-dimethyl-1h-pyrazole-5-carboxylic acid, 1,3-dimethylpyrazole-5-carboxylic acid, 2,5-dimethyl-2h-pyrazole-3-carboxylic acid, 1h-pyrazole-5-carboxylic acid, 1,3-dimethyl, 1,3-dimethylpyrazole-5-carboxylicacid, pubchem7839, ksc269g2l, 1,3-dimethylpyrazol-5-carboxylic acid, pyrazole-5-carboxylic acid, 1,3-dmethyl, 1h-pyrazole-5-carboxylicacid, 1,3-dimethyl
PubChem CID 587721
IUPAC Name 2,5-dimethylpyrazole-3-carboxylic acid
SMILES CN1N=C(C)C=C1C(O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC12501DA
SDS View Documents
Thermo Scientific Chemicals
CC12501DA
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC12501DE
SDS View Documents
Thermo Scientific Chemicals
CC12501DE
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC12501EA
SDS View Documents
Thermo Scientific Chemicals
CC12501EA
10g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

5744-56-9
140.14
QRWZFUBHOQWUGH-UHFFFAOYSA-N
587721
CN1N=C(C)C=C1C(O)=O
C6H8N2O2
MFCD00159636
1,3-dimethyl-1h-pyrazole-5-carboxylic acid, 1,3-dimethylpyrazole-5-carboxylic acid, 2,5-dimethyl-2h-pyrazole-3-carboxylic acid, 1h-pyrazole-5-carboxylic acid, 1,3-dimethyl, 1,3-dimethylpyrazole-5-carboxylicacid, pubchem7839, ksc269g2l, 1,3-dimethylpyrazol-5-carboxylic acid, pyrazole-5-carboxylic acid, 1,3-dmethyl, 1h-pyrazole-5-carboxylicacid, 1,3-dimethyl
2,5-dimethylpyrazole-3-carboxylic acid

Specifications

5744-56-9
C6H8N2O2
1g
QRWZFUBHOQWUGH-UHFFFAOYSA-N
2,5-dimethylpyrazole-3-carboxylic acid
587721
97%
97%
MFCD00159636
1,3-dimethyl-1h-pyrazole-5-carboxylic acid, 1,3-dimethylpyrazole-5-carboxylic acid, 2,5-dimethyl-2h-pyrazole-3-carboxylic acid, 1h-pyrazole-5-carboxylic acid, 1,3-dimethyl, 1,3-dimethylpyrazole-5-carboxylicacid, pubchem7839, ksc269g2l, 1,3-dimethylpyrazol-5-carboxylic acid, pyrazole-5-carboxylic acid, 1,3-dmethyl, 1h-pyrazole-5-carboxylicacid, 1,3-dimethyl
CN1N=C(C)C=C1C(O)=O
140.14
140.14
1, 3-Dimethyl-1H-pyrazole-5-carboxylic acid
Videos
Safety and Handling

Safety and Handling

GHS P Statement Causes skin irritation. Causes serious eye irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.

Warning

SDS
missing translation for 'documents'

missing translation for 'documents'