missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3-Dimethyl-3,4,5,6-tetrahydro-2(1H)-pyrimidinone, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L0996822

 View more versions of this product

Catalog No. AAL0996822


Description

Description

1,3-Dimethyl-3,4,5,6-tetrahydro-2(1H)-pyrimidinone is a versatile solvent used in the N-alkylation of chiral and O-alkylation of aldoses. It is involved in the preparation of poly(aryl ethers). It is a cyclic urea and used as a polar aprotic organic solvent. Further, it reacts with trifluoro-methanesulfonic acid anhydride to prepare 2,2'-oxy-bis(1,3-dimethyl-tetrahydropyrimidinium) bis(trifluoromethanesulfonate).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Dimethyl-3,4,5,6-tetrahydro-2(1H)-pyrimidinone is a versatile solvent used in the N-alkylation of chiral and O-alkylation of aldoses. It is involved in the preparation of poly(aryl ethers). It is a cyclic urea and used as a polar aprotic organic solvent. Further, it reacts with trifluoro-methanesulfonic acid anhydride to prepare 2,2′-oxy-bis(1,3-dimethyl-tetrahydropyrimidinium) bis(trifluoromethanesulfonate).

Solubility
Miscible with water.

Notes
Hygroscopic. Moisture sensitive. Incompatible with strong oxidizing agents.
Specifications

Specifications

1,3-Dimethyl-3,4,5,6-tetrahydro-2(1H)-pyrimidinone
-23°C
11
120°C (248°F)
C6H12N2O
MFCD00006550
110562
1,3-dimethyl-3,4,5,6-tetrahydro-2 1h-pyrimidinone, 1,3-dimethyltetrahydropyrimidin-2 1h-one, dmpu, dimethylpropyleneurea, 2 1h-pyrimidinone, tetrahydro-1,3-dimethyl, n,n'-dimethylpropyleneurea, n,n'-dimethyltrimethyleneurea, ccris 4322, n,n'-dimethylpropylene urea, tetrahydro-1,3-dimethyl-2 1h pyrimidine
GUVUOGQBMYCBQP-UHFFFAOYSA-N
1,3-dimethyl-1,3-diazinan-2-one
81646
98%
7226-23-5
1.06
246°C to 247°C
Odorless
1.489
100 g
Hygroscopic
Miscible with water.
CN1CCCN(C1=O)C
128.175
128.18

Chemical Identifiers

7226-23-5
128.175
GUVUOGQBMYCBQP-UHFFFAOYSA-N
81646
CN1CCCN(C1=O)C
C6H12N2O
MFCD00006550
1,3-dimethyl-3,4,5,6-tetrahydro-2 1h-pyrimidinone, 1,3-dimethyltetrahydropyrimidin-2 1h-one, dmpu, dimethylpropyleneurea, 2 1h-pyrimidinone, tetrahydro-1,3-dimethyl, n,n'-dimethylpropyleneurea, n,n'-dimethyltrimethyleneurea, ccris 4322, n,n'-dimethylpropylene urea, tetrahydro-1,3-dimethyl-2 1h pyrimidine
1,3-dimethyl-1,3-diazinan-2-one
Safety and Handling

Safety and Handling

GHS H Statement
H318-H361-H302
Causes serious eye damage.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.

P201-P202-P264b-P270-P281-P301+P312-P305+P351+P338-P308+P313-P310-P330-P501c

H302-H318-H361f

EINECSNumber : 230-625-6

RTECSNumber : UW7582500

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only