missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide 98.0+%, TCI America™
Supplier: TCI America D402925G
Specifications
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide | |
Yellow | |
C8H17N3 | |
25 g | |
1-3-dimethylaminopropyl-3-ethylcarbodiimide, edac, 1-ethyl-3-3-dimethylaminopropyl carbodiimide, n1-ethylimino methylene-n3,n3-dimethylpropane-1,3-diamine, unii-rj5ozg6i4a, edci, 1,3-propanediamine, n'-ethylcarbonimidoyl-n,n-dimethyl, n-3-dimethylaminopropyl-n'-ethylcarbodiimide, rj5ozg6i4a, ethyldimethylaminopropyl carbodiimide | |
CCN=C=NCCCN(C)C | |
155.245 | |
155.25 | |
Liquid |
1892-57-5 | |
45°C | |
MFCD00044916 | |
2735 | |
LMDZBCPBFSXMTL-UHFFFAOYSA-N | |
3-(ethyliminomethylideneamino)-N,N-dimethylpropan-1-amine | |
15908 | |
≥98.0% (GC,T) |
Chemical Identifiers
1892-57-5 | |
155.245 | |
LMDZBCPBFSXMTL-UHFFFAOYSA-N | |
15908 | |
CCN=C=NCCCN(C)C |
C8H17N3 | |
MFCD00044916 | |
1-3-dimethylaminopropyl-3-ethylcarbodiimide, edac, 1-ethyl-3-3-dimethylaminopropyl carbodiimide, n1-ethylimino methylene-n3,n3-dimethylpropane-1,3-diamine, unii-rj5ozg6i4a, edci, 1,3-propanediamine, n'-ethylcarbonimidoyl-n,n-dimethyl, n-3-dimethylaminopropyl-n'-ethylcarbodiimide, rj5ozg6i4a, ethyldimethylaminopropyl carbodiimide | |
3-(ethyliminomethylideneamino)-N,N-dimethylpropan-1-amine |
Safety and Handling
EINECSNumber : (2)-1696
TSCA : Yes
Recommended Storage : Freezer