missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,3-Dimethylcyclohexane (cis- and trans- mixture) 97.0+%, TCI America™
$108.62 - $809.75
Chemical Identifiers
CAS | 591-21-9 |
---|---|
Molecular Formula | C8H16 |
Molecular Weight (g/mol) | 112.216 |
MDL Number | MFCD00064173 |
InChI Key | SGVUHPSBDNVHKL-UHFFFAOYSA-N |
Synonym | hexahydro-m-xylene, cyclohexane, 1,3-dimethyl, 1,3-dimethylcyclohexane,c&t, trans-1,3-dimethylcyclohexane, 1,3-dimethylcyclohexane, cis + trans, unii-9sgo12qv70, 1,3-dimethylcyclohexane, cis, m-dimethylcyclohexane, acmc-209nia, 1,3-dimethyl cyclohexane |
PubChem CID | 11564 |
IUPAC Name | 1,3-dimethylcyclohexane |
SMILES | CC1CCCC(C1)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
D069925ML
|
TCI America
D069925ML |
25 mL |
Each for $108.62
|
|
|||||
D0699500ML
|
TCI America
D0699500ML |
500 mL |
Each for $809.75
|
|
|||||
Chemical Identifiers
591-21-9 | |
112.216 | |
SGVUHPSBDNVHKL-UHFFFAOYSA-N | |
11564 | |
CC1CCCC(C1)C |
C8H16 | |
MFCD00064173 | |
hexahydro-m-xylene, cyclohexane, 1,3-dimethyl, 1,3-dimethylcyclohexane,c&t, trans-1,3-dimethylcyclohexane, 1,3-dimethylcyclohexane, cis + trans, unii-9sgo12qv70, 1,3-dimethylcyclohexane, cis, m-dimethylcyclohexane, acmc-209nia, 1,3-dimethyl cyclohexane | |
1,3-dimethylcyclohexane |
Specifications
591-21-9 | |
C8H16 | |
25 mL | |
hexahydro-m-xylene, cyclohexane, 1,3-dimethyl, 1,3-dimethylcyclohexane,c&t, trans-1,3-dimethylcyclohexane, 1,3-dimethylcyclohexane, cis + trans, unii-9sgo12qv70, 1,3-dimethylcyclohexane, cis, m-dimethylcyclohexane, acmc-209nia, 1,3-dimethyl cyclohexane | |
CC1CCCC(C1)C | |
112.216 | |
112.22 | |
Liquid |
Colorless | |
MFCD00064173 | |
2263 | |
SGVUHPSBDNVHKL-UHFFFAOYSA-N | |
1,3-dimethylcyclohexane | |
11564 | |
≥97.0% (GC) | |
1,3-Dimethylcyclohexane (cis- and trans- mixture) |
Safety and Handling
EINECSNumber : (3)-2227
TSCA : No