missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,3-Diphenylacetone, 98+%, Thermo Scientific Chemicals
$93.92 - $332.44
Chemical Identifiers
CAS | 102-04-5 |
---|---|
Molecular Formula | C15H14O |
Molecular Weight (g/mol) | 210.28 |
MDL Number | MFCD00004795 |
InChI Key | YFKBXYGUSOXJGS-UHFFFAOYSA-N |
Synonym | 1,3-diphenylacetone, dibenzyl ketone, 1,3-diphenyl-2-propanone, benzyl ketone, 2-propanone, 1,3-diphenyl, 1,3-diphenylpropanone, alpha,alpha'-diphenylacetone, 1,3-diphenyl-propan-2-one, unii-9y07g5udkq, fema no. 2397 |
PubChem CID | 7593 |
IUPAC Name | 1,3-diphenylpropan-2-one |
SMILES | O=C(CC1=CC=CC=C1)CC1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0283014
|
Thermo Scientific Chemicals
L0283014 |
25 g |
Each for $93.92
|
|
|||||
AAL0283022
|
Thermo Scientific Chemicals
L0283022 |
100 g |
Each for $332.44
|
|
|||||
Description
1,3-Diphenylacetone is used in the aldol condensation reaction with benzil (a dicarbonyl) and base to create tetraphenylcyclopentadienone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,3-Diphenylacetone is used in the aldol condensation reaction with benzil (a dicarbonyl) and base to create tetraphenylcyclopentadienone.
Solubility
Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
102-04-5 | |
210.28 | |
YFKBXYGUSOXJGS-UHFFFAOYSA-N | |
7593 | |
O=C(CC1=CC=CC=C1)CC1=CC=CC=C1 |
C15H14O | |
MFCD00004795 | |
1,3-diphenylacetone, dibenzyl ketone, 1,3-diphenyl-2-propanone, benzyl ketone, 2-propanone, 1,3-diphenyl, 1,3-diphenylpropanone, alpha,alpha'-diphenylacetone, 1,3-diphenyl-propan-2-one, unii-9y07g5udkq, fema no. 2397 | |
1,3-diphenylpropan-2-one |
Specifications
102-04-5 | |
1.04 | |
>110°C (230°F) | |
MFCD00004795 | |
974767 | |
Slightly soluble in water. | |
O=C(CC1=CC=CC=C1)CC1=CC=CC=C1 | |
210.28 | |
210.28 | |
1,3-Diphenylacetone |
32°C to 36°C | |
330°C | |
C15H14O | |
25 g | |
1,3-diphenylacetone, dibenzyl ketone, 1,3-diphenyl-2-propanone, benzyl ketone, 2-propanone, 1,3-diphenyl, 1,3-diphenylpropanone, alpha,alpha'-diphenylacetone, 1,3-diphenyl-propan-2-one, unii-9y07g5udkq, fema no. 2397 | |
YFKBXYGUSOXJGS-UHFFFAOYSA-N | |
1,3-diphenylpropan-2-one | |
7593 | |
≥98% |
Safety and Handling
EINECSNumber : 203-000-0
RTECSNumber : UC2010030
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only