missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3-Dithiane, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1050522

 View more versions of this product

Catalog No. AAA1050522


Explore available promotions

Description

Description

1,3-Dithiane is used in umpolung reactions such as the Corey-Seebach reaction. It acts as a reagent for the deoxygenation of sulfoxides to their corresponding sulfides. Further, it is used as a protecting group for carbonyl compounds used in organic synthesis. It serves as a useful labeled synthon. In addition to this, it is used in the preparation of 2-ethyl-[1,3]dithiane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Dithiane is used in umpolung reactions such as the Corey-Seebach reaction. It acts as a reagent for the deoxygenation of sulfoxides to their corresponding sulfides. Further, it is used as a protecting group for carbonyl compounds used in organic synthesis. It serves as a useful labeled synthon. In addition to this, it is used in the preparation of 2-ethyl-[1,3]dithiane.

Solubility
Soluble in benzene, ether, chloroform and terahydrofuran. Slightly soluble in water.

Notes
Hygroscopic. Incompatible with strong bases and strong oxidizing agents.
Specifications

Specifications

1,3-Dithiane
52°C to 55°C
90°C (194°F)
C4H8S2
100 g
102534
Soluble in benzene,ether,chloroform and terahydrofuran. Slightly soluble in water.
C1CSCSC1
120.228
CHEBI:47845
98%
505-23-7
207°C to 208°C
Stench
MFCD00006654
UN3335
1,3-dithiacyclohexane, m-dithiane, dithiane-1,3, 1,3-dithian, dithiane-1,3 french, 1,3 dithiane, ccris 6777, m-dithiane, 8ci, 1,3 dithiocyclohexane, acmc-209knk
WQADWIOXOXRPLN-UHFFFAOYSA-N
1,3-dithiane
10451
120.24

Chemical Identifiers

505-23-7
120.228
WQADWIOXOXRPLN-UHFFFAOYSA-N
10451
1,3-dithiane
C4H8S2
MFCD00006654
1,3-dithiacyclohexane, m-dithiane, dithiane-1,3, 1,3-dithian, dithiane-1,3 french, 1,3 dithiane, ccris 6777, m-dithiane, 8ci, 1,3 dithiocyclohexane, acmc-209knk
CHEBI:47845
C1CSCSC1
Videos
Safety and Handling

Safety and Handling

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: AVIATION REGULATED SOLID, N.O.S.

EINECSNumber : 208-006-7

RTECSNumber : JO5070000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only