missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(3-Hydroxybenzyl)piperidine 98.0+%, TCI America™
$195.73 - $777.32
Chemical Identifiers
| CAS | 73279-04-6 |
|---|---|
| Molecular Formula | C12H17NO |
| Molecular Weight (g/mol) | 191.274 |
| MDL Number | MFCD00800250 |
| InChI Key | ORGBERFQYFWYGX-UHFFFAOYSA-N |
| Synonym | 3-1-piperidinylmethyl phenol, 3-piperidin-1-ylmethyl phenol, 3-1-piperidinomethyl phenol, 1-3-hydroxybenzyl piperidine, 3-piperidinomethyl phenol, 3-1-piperidylmethyl phenol, phenol, 3-1-piperidinylmethyl, 1-3-hydroxyphenylmethyl piperidine, pubchem6815, 3-piperidinomethylphenol |
| PubChem CID | 826974 |
| IUPAC Name | 3-(piperidin-1-ylmethyl)phenol |
| SMILES | C1CCN(CC1)CC2=CC(=CC=C2)O |
Chemical Identifiers
| 73279-04-6 | |
| 191.274 | |
| ORGBERFQYFWYGX-UHFFFAOYSA-N | |
| 826974 | |
| C1CCN(CC1)CC2=CC(=CC=C2)O |
| C12H17NO | |
| MFCD00800250 | |
| 3-1-piperidinylmethyl phenol, 3-piperidin-1-ylmethyl phenol, 3-1-piperidinomethyl phenol, 1-3-hydroxybenzyl piperidine, 3-piperidinomethyl phenol, 3-1-piperidylmethyl phenol, phenol, 3-1-piperidinylmethyl, 1-3-hydroxyphenylmethyl piperidine, pubchem6815, 3-piperidinomethylphenol | |
| 3-(piperidin-1-ylmethyl)phenol |
Specifications
| 73279-04-6 | |
| White-Yellow | |
| MFCD00800250 | |
| 3-1-piperidinylmethyl phenol, 3-piperidin-1-ylmethyl phenol, 3-1-piperidinomethyl phenol, 1-3-hydroxybenzyl piperidine, 3-piperidinomethyl phenol, 3-1-piperidylmethyl phenol, phenol, 3-1-piperidinylmethyl, 1-3-hydroxyphenylmethyl piperidine, pubchem6815, 3-piperidinomethylphenol | |
| C1CCN(CC1)CC2=CC(=CC=C2)O | |
| 191.274 | |
| 191.27 | |
| Crystalline Powder |
| 139°C | |
| C12H17NO | |
| 5 g | |
| ORGBERFQYFWYGX-UHFFFAOYSA-N | |
| 3-(piperidin-1-ylmethyl)phenol | |
| 826974 | |
| ≥98.0% (GC) | |
| 1-(3-Hydroxybenzyl)piperidine |
Safety and Handling
TSCA : No