missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Phenylenediamine-4-sulfonic Acid 98.0+%, TCI America™
$74.56 - $622.59
Chemical Identifiers
| CAS | 88-63-1 |
|---|---|
| Molecular Formula | C6H8N2O3S |
| Molecular Weight (g/mol) | 188.201 |
| MDL Number | MFCD00035772 |
| InChI Key | JVMSQRAXNZPDHF-UHFFFAOYSA-N |
| Synonym | 2,4-Diaminobenzenesulfonic Acid |
| PubChem CID | 66623 |
| IUPAC Name | 2,4-diaminobenzenesulfonic acid |
| SMILES | C1=CC(=C(C=C1N)N)S(=O)(=O)O |
Chemical Identifiers
| 88-63-1 | |
| 188.201 | |
| JVMSQRAXNZPDHF-UHFFFAOYSA-N | |
| 66623 | |
| C1=CC(=C(C=C1N)N)S(=O)(=O)O |
| C6H8N2O3S | |
| MFCD00035772 | |
| 2,4-Diaminobenzenesulfonic Acid | |
| 2,4-diaminobenzenesulfonic acid |
Specifications
| 88-63-1 | |
| C6H8N2O3S | |
| 25 g | |
| JVMSQRAXNZPDHF-UHFFFAOYSA-N | |
| 2,4-diaminobenzenesulfonic acid | |
| 66623 | |
| ≥98.0% (HPLC,T) | |
| 1,3-Phenylenediamine-4-sulfonic Acid |
| Yellow | |
| MFCD00035772 | |
| 2,4-Diaminobenzenesulfonic Acid | |
| C1=CC(=C(C=C1N)N)S(=O)(=O)O | |
| 188.201 | |
| 188.20 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-2058
RTECSNumber : DB5900000
TSCA : Yes