Learn More
1,3-Propanesultone, 99%, Thermo Scientific Chemicals
$76.86 - $567.11
Chemical Identifiers
CAS | 1120-71-4 |
---|---|
Molecular Formula | C3H6O3S |
Molecular Weight (g/mol) | 122.14 |
MDL Number | MFCD00005355 |
InChI Key | FSSPGSAQUIYDCN-UHFFFAOYSA-N |
Synonym | 1,3-propanesultone, propane sultone, 1,2-oxathiolane 2,2-dioxide, 1,3-propane sultone, 1,2-oxathiolane, 2,2-dioxide, propanesultone, rcra waste number u193, .gamma.-propane sultone, 3-hydroxy-1-propanesulfonic acid gamma-sultone, unii-l6ntk7vjx9 |
PubChem CID | 14264 |
ChEBI | CHEBI:82370 |
SMILES | O=S1(=O)CCCO1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1192309
|
Thermo Scientific Chemicals
A1192309 |
10 g |
Each for $76.86
|
|
|||||
AAA1192318
|
Thermo Scientific Chemicals
A1192318 |
50 g |
Each for $168.62
|
|
|||||
AAA1192330
|
Thermo Scientific Chemicals
A1192330 |
250 g |
Each for $567.11
|
|
|||||
Description
1,3-Propane sultone is used as a chemical intermediate to introduce sulfopropyl groups into molecules and to confer water solubility and anionic character.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,3-Propane sultone is used as a chemical intermediate to introduce sulfopropyl groups into molecules and to confer water solubility and anionic character.
Solubility
Soluble in water (100 mg/ml), chloroform, esters, methanol, and aromatic hydrocartions.
Notes
Special handling precautions required. View MSDS prior to purchase. MSDS are available online at www.alfa.com
Chemical Identifiers
1120-71-4 | |
122.14 | |
FSSPGSAQUIYDCN-UHFFFAOYSA-N | |
14264 | |
O=S1(=O)CCCO1 |
C3H6O3S | |
MFCD00005355 | |
1,3-propanesultone, propane sultone, 1,2-oxathiolane 2,2-dioxide, 1,3-propane sultone, 1,2-oxathiolane, 2,2-dioxide, propanesultone, rcra waste number u193, .gamma.-propane sultone, 3-hydroxy-1-propanesulfonic acid gamma-sultone, unii-l6ntk7vjx9 | |
CHEBI:82370 |
Specifications
1120-71-4 | |
1.392 | |
>110°C (230°F) | |
C3H6O3S | |
10 g | |
Moisture sensitive | |
Soluble in water (100mg/ml),chloroform,esters,methanol,and aromatic hydrocartions. | |
O=S1(=O)CCCO1 | |
122.14 | |
CHEBI:82370 | |
1, 3-Propanesultone |
30°C to 33°C | |
179°C to 181°C (30 mm) | |
99% | |
MFCD00005355 | |
109782 | |
1,3-propanesultone, propane sultone, 1,2-oxathiolane 2,2-dioxide, 1,3-propane sultone, 1,2-oxathiolane, 2,2-dioxide, propanesultone, rcra waste number u193, .gamma.-propane sultone, 3-hydroxy-1-propanesulfonic acid gamma-sultone, unii-l6ntk7vjx9 | |
FSSPGSAQUIYDCN-UHFFFAOYSA-N | |
oxathiolane 2,2-dioxide | |
14264 | |
122.14 |
Safety and Handling
P201-P202-P264b-P270-P281-P301+P312-P302+P352-P308+P313-P312-P330-P363-P501c
H302+H312-H350
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 214-317-9
RTECSNumber : RP5425000
TSCA : Yes
storageNote1 : Moisture Sensitive
Recommended Storage : Ambient temperatures
RUO – Research Use Only