Learn More
1,3-Thiazole-2-carbonyl chloride, ≥90%, Thermo Scientific™
$194.26 - $194.26
Chemical Identifiers
CAS | 30216-57-0 |
---|---|
Molecular Formula | C4H2ClNOS |
Molecular Weight (g/mol) | 147.576 |
MDL Number | MFCD06658970 |
InChI Key | HKZCFGQSWLWNQA-UHFFFAOYSA-N |
Synonym | thiazole-2-carbonyl chloride, 2-thiazolecarbonylchloride, 2-thiazolecarbonyl chloride, thiazole carboxylic acid chloride, 1,3-thiazole-2carbonyl chloride, 1,3-thiazole-2-carbonyl-chloride, 2-chlorocarbonyl-1,3-thiazole, 2-thiazolecarbonyl chloride 8ci,9ci |
PubChem CID | 2795211 |
IUPAC Name | 1,3-thiazole-2-carbonyl chloride |
SMILES | C1=CSC(=N1)C(=O)Cl |
Chemical Identifiers
30216-57-0 | |
147.576 | |
HKZCFGQSWLWNQA-UHFFFAOYSA-N | |
2795211 | |
C1=CSC(=N1)C(=O)Cl |
C4H2ClNOS | |
MFCD06658970 | |
thiazole-2-carbonyl chloride, 2-thiazolecarbonylchloride, 2-thiazolecarbonyl chloride, thiazole carboxylic acid chloride, 1,3-thiazole-2carbonyl chloride, 1,3-thiazole-2-carbonyl-chloride, 2-chlorocarbonyl-1,3-thiazole, 2-thiazolecarbonyl chloride 8ci,9ci | |
1,3-thiazole-2-carbonyl chloride |
Specifications
30216-57-0 | |
100.0 | |
Amber Glass Bottle | |
MFCD06658970 | |
thiazole-2-carbonyl chloride, 2-thiazolecarbonylchloride, 2-thiazolecarbonyl chloride, thiazole carboxylic acid chloride, 1,3-thiazole-2carbonyl chloride, 1,3-thiazole-2-carbonyl-chloride, 2-chlorocarbonyl-1,3-thiazole, 2-thiazolecarbonyl chloride 8ci,9ci | |
C1=CSC(=N1)C(=O)Cl | |
147.576 | |
147.58 | |
1, 3-Thiazole-2-carbonyl chloride |
89.5 | |
90% | |
C4H2ClNOS | |
250 mg | |
HKZCFGQSWLWNQA-UHFFFAOYSA-N | |
1,3-thiazole-2-carbonyl chloride | |
2795211 | |
90% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger