Learn More
1-(3-Trifluoromethylphenoxy)-3-butyn-2-ol, 98%, Thermo Scientific Chemicals
$106.47 - $308.63
Chemical Identifiers
CAS | 88462-65-1 |
---|---|
Molecular Formula | C11H9F3O2 |
Molecular Weight (g/mol) | 230.19 |
MDL Number | MFCD02683085 |
InChI Key | OPWHACCQYXGLRX-UHFFFAOYNA-N |
Synonym | 1-3-trifluoromethylphenoxy-3-butyn-2-ol, 1-3-trifluoromethyl phenoxy but-3-yn-2-ol, 3-butyn-2-ol,1-3-trifluoromethyl phenoxy, acmc-20drht, 1-3-trifluoromethylphenoxy-3-butyn-2-o |
PubChem CID | 11172256 |
IUPAC Name | 1-[3-(trifluoromethyl)phenoxy]but-3-yn-2-ol |
SMILES | OC(COC1=CC=CC(=C1)C(F)(F)F)C#C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAB2221803
|
Thermo Scientific Chemicals
B2221803 |
1 g |
Each for $106.47
|
|
||||||
AAB2221806
|
Thermo Scientific Chemicals
B2221806 |
5 g |
Each for $308.63
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
88462-65-1 | |
230.19 | |
OPWHACCQYXGLRX-UHFFFAOYNA-N | |
11172256 | |
OC(COC1=CC=CC(=C1)C(F)(F)F)C#C |
C11H9F3O2 | |
MFCD02683085 | |
1-3-trifluoromethylphenoxy-3-butyn-2-ol, 1-3-trifluoromethyl phenoxy but-3-yn-2-ol, 3-butyn-2-ol,1-3-trifluoromethyl phenoxy, acmc-20drht, 1-3-trifluoromethylphenoxy-3-butyn-2-o | |
1-[3-(trifluoromethyl)phenoxy]but-3-yn-2-ol |
Specifications
88462-65-1 | |
C11H9F3O2 | |
1 g | |
OPWHACCQYXGLRX-UHFFFAOYNA-N | |
1-[3-(trifluoromethyl)phenoxy]but-3-yn-2-ol | |
11172256 | |
98% |
30°C to 33°C | |
MFCD02683085 | |
1-3-trifluoromethylphenoxy-3-butyn-2-ol, 1-3-trifluoromethyl phenoxy but-3-yn-2-ol, 3-butyn-2-ol,1-3-trifluoromethyl phenoxy, acmc-20drht, 1-3-trifluoromethylphenoxy-3-butyn-2-o | |
OC(COC1=CC=CC(=C1)C(F)(F)F)C#C | |
230.19 | |
230.19 | |
1-(3-Trifluoromethylphenoxy)-3-butyn-2-ol |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only