missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-[4-(3-Bromothien-2-yl)phenyl]ethanone, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC67828DA

 View more versions of this product

Catalog No. CC67828DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

1-[4-(3-Bromothien-2-yl)phenyl]ethanone
96.5
97%
C12H9BrOS
1g
AMADTVLVNSCDKW-UHFFFAOYSA-N
1-[4-(3-bromothiophen-2-yl)phenyl]ethanone
24229756
97%
937796-01-5
100.0
Amber Glass Bottle
MFCD09879973
1-4-3-bromothien-2-yl phenyl ethanone, 1-4-3-bromothiophen-2-yl phenyl ethanone, 1-4-3-bromothiophen-2-yl phenyl ethan-1-one, 4'-3-bromothien-2-yl acetophenone
CC(=O)C1=CC=C(C=C1)C2=C(C=CS2)Br
281.167
281.17

Chemical Identifiers

937796-01-5
281.167
AMADTVLVNSCDKW-UHFFFAOYSA-N
24229756
CC(=O)C1=CC=C(C=C1)C2=C(C=CS2)Br
C12H9BrOS
MFCD09879973
1-4-3-bromothien-2-yl phenyl ethanone, 1-4-3-bromothiophen-2-yl phenyl ethanone, 1-4-3-bromothiophen-2-yl phenyl ethan-1-one, 4'-3-bromothien-2-yl acetophenone
1-[4-(3-bromothiophen-2-yl)phenyl]ethanone
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions