missing translation for 'onlineSavingsMsg'
Learn More

1,4-Benzodioxan-6-amine, 99%

CAS: 22013-33-8 | C8H9NO2 | 151.17 g/mol

$392.54 - $392.54

Chemical Identifiers

CAS 22013-33-8
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00006824
InChI Key BZKOZYWGZKRTIB-UHFFFAOYSA-N
Synonym 1,4-benzodioxan-6-amine, 3,4-ethylenedioxyaniline, 6-amino-1,4-benzodioxane, 2,3-dihydrobenzo b 1,4 dioxin-6-amine, 6-amino-1,4-benzodioxan, 1,4-benzodioxan-6-ylamine, 1,4-benzodioxin-6-amine, 2,3-dihydro, 3,4-ethylenedioxy aniline, 2,3-dihydro-benzo 1,4 dioxin-6-ylamine, 2,3-dihydro-1,4-benzodioxin-6-amin
PubChem CID 89148
IUPAC Name 2,3-dihydro-1,4-benzodioxin-6-amine
SMILES NC1=CC=C2OCCOC2=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC185780500
SDS View Documents
Thermo Scientific Chemicals
185780500
50 g Glass bottle
Each for $392.54
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

22013-33-8
151.17
BZKOZYWGZKRTIB-UHFFFAOYSA-N
89148
NC1=CC=C2OCCOC2=C1
C8H9NO2
MFCD00006824
1,4-benzodioxan-6-amine, 3,4-ethylenedioxyaniline, 6-amino-1,4-benzodioxane, 2,3-dihydrobenzo b 1,4 dioxin-6-amine, 6-amino-1,4-benzodioxan, 1,4-benzodioxan-6-ylamine, 1,4-benzodioxin-6-amine, 2,3-dihydro, 3,4-ethylenedioxy aniline, 2,3-dihydro-benzo 1,4 dioxin-6-ylamine, 2,3-dihydro-1,4-benzodioxin-6-amin
2,3-dihydro-1,4-benzodioxin-6-amine

Specifications

22013-33-8
100.0
1.2300g/mL
Authentic
Glass bottle
1.5977 to 1.5997
50 g
1,4-benzodioxan-6-amine, 3,4-ethylenedioxyaniline, 6-amino-1,4-benzodioxane, 2,3-dihydrobenzo b 1,4 dioxin-6-amine, 6-amino-1,4-benzodioxan, 1,4-benzodioxan-6-ylamine, 1,4-benzodioxin-6-amine, 2,3-dihydro, 3,4-ethylenedioxy aniline, 2,3-dihydro-benzo 1,4 dioxin-6-ylamine, 2,3-dihydro-1,4-benzodioxin-6-amin
BZKOZYWGZKRTIB-UHFFFAOYSA-N
2,3-dihydro-1,4-benzodioxin-6-amine
89148
99%
1, 4-Benzodioxan-6-amine
98.5
Brown to Black
120°C
99%
C8H9NO2
MFCD00006824
1.23
Solubility in water: insoluble. Other solubilities: soluble in many organic solvents
NC1=CC=C2OCCOC2=C1
151.17
151.16
Viscous Liquid After Melting
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 244-718-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only