Learn More
1,4'-Bipiperidine, 99%, Thermo Scientific Chemicals
$94.85 - $649.93
Chemical Identifiers
CAS | 4897-50-1 |
---|---|
Molecular Formula | C10H20N2 |
Molecular Weight (g/mol) | 168.284 |
MDL Number | MFCD00006475 |
InChI Key | QDVBKXJMLILLLB-UHFFFAOYSA-N |
Synonym | 4-piperidinopiperidine, 1,4'-bipiperidine, 4-piperidino-piperidine, 1,4'-bipiperidyl, 4-1-piperidinyl piperidine, 1,4' bipiperidinyl, 4-piperidin-1-yl piperidine, 1,4-bipiperidine, 1-piperidin-4-yl piperidine, 4-piperidin-1-ylpiperidine |
PubChem CID | 78607 |
ChEBI | CHEBI:40117 |
IUPAC Name | 1-piperidin-4-ylpiperidine |
SMILES | C1CCN(CC1)C2CCNCC2 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2202803
|
Thermo Scientific Chemicals
B2202803 |
1 g |
Each for $94.85
|
|
|||||
AAB2202806
|
Thermo Scientific Chemicals
B2202806 |
5 g |
Each for $197.18
|
|
|||||
AAB2202814
|
Thermo Scientific Chemicals
B2202814 |
25 g |
Each for $649.93
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
4897-50-1 | |
168.284 | |
QDVBKXJMLILLLB-UHFFFAOYSA-N | |
78607 | |
1-piperidin-4-ylpiperidine |
C10H20N2 | |
MFCD00006475 | |
4-piperidinopiperidine, 1,4'-bipiperidine, 4-piperidino-piperidine, 1,4'-bipiperidyl, 4-1-piperidinyl piperidine, 1,4' bipiperidinyl, 4-piperidin-1-yl piperidine, 1,4-bipiperidine, 1-piperidin-4-yl piperidine, 4-piperidin-1-ylpiperidine | |
CHEBI:40117 | |
C1CCN(CC1)C2CCNCC2 |
Specifications
4897-50-1 | |
C10H20N2 | |
1 g | |
Air Sensitive | |
QDVBKXJMLILLLB-UHFFFAOYSA-N | |
1-piperidin-4-ylpiperidine | |
78607 | |
168.28 | |
1,4'-Bipiperidine |
66°C to 70°C | |
MFCD00006475 | |
UN3259 | |
4-piperidinopiperidine, 1,4'-bipiperidine, 4-piperidino-piperidine, 1,4'-bipiperidyl, 4-1-piperidinyl piperidine, 1,4' bipiperidinyl, 4-piperidin-1-yl piperidine, 1,4-bipiperidine, 1-piperidin-4-yl piperidine, 4-piperidin-1-ylpiperidine | |
C1CCN(CC1)C2CCNCC2 | |
168.284 | |
CHEBI:40117 | |
99% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
EINECSNumber : 225-522-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only