Learn More
1,4-Bis(2-hydroxyisopropyl)benzene, 97%, Thermo Scientific Chemicals
$55.05 - $162.84
Chemical Identifiers
CAS | 2948-46-1 |
---|---|
Molecular Formula | C12H18O2 |
Molecular Weight (g/mol) | 194.274 |
MDL Number | MFCD00009827 |
InChI Key | LEARFTRDZQQTDN-UHFFFAOYSA-N |
Synonym | 1,4-bis 2-hydroxy-2-propyl benzene, 1,4-bis 1-methyl-1-hydroxyethyl benzene, alpha,alpha,alpha',alpha'-tetramethyl-1,4-benzenedimethanol, unii-g785drb5xh, p-bis 2-hydroxyisopropyl benzene, 1,4-bis 2-hydroxyisopropyl benzene, p-bis alpha-hydroxyisopropyl benzene, g785drb5xh, alpha,alpha'-dihydroxy-p-diisopropylbenzene, alpha,alpha,alpha',alpha'-tetramethyl-p-xylylenediol |
PubChem CID | 18062 |
IUPAC Name | 2-[4-(2-hydroxypropan-2-yl)phenyl]propan-2-ol |
SMILES | CC(C)(C1=CC=C(C=C1)C(C)(C)O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2147203
|
Thermo Scientific Chemicals
B2147203 |
1 g |
Each for $55.05
|
|
|||||
AAB2147206
|
Thermo Scientific Chemicals
B2147206 |
5 g |
Each for $162.84
|
|
|||||
Description
1,4-Bis(2-hydroxyisopropyl)benzene is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,4-Bis(2-hydroxyisopropyl)benzene is used as a pharmaceutical intermediate.
Solubility
Slightly Soluble in water (2.1 g/L) (25°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from acids, acid chlorides, acid anhydrides, strong oxidizing agents.
Chemical Identifiers
2948-46-1 | |
194.274 | |
LEARFTRDZQQTDN-UHFFFAOYSA-N | |
18062 | |
CC(C)(C1=CC=C(C=C1)C(C)(C)O)O |
C12H18O2 | |
MFCD00009827 | |
1,4-bis 2-hydroxy-2-propyl benzene, 1,4-bis 1-methyl-1-hydroxyethyl benzene, alpha,alpha,alpha',alpha'-tetramethyl-1,4-benzenedimethanol, unii-g785drb5xh, p-bis 2-hydroxyisopropyl benzene, 1,4-bis 2-hydroxyisopropyl benzene, p-bis alpha-hydroxyisopropyl benzene, g785drb5xh, alpha,alpha'-dihydroxy-p-diisopropylbenzene, alpha,alpha,alpha',alpha'-tetramethyl-p-xylylenediol | |
2-[4-(2-hydroxypropan-2-yl)phenyl]propan-2-ol |
Specifications
2948-46-1 | |
167°C to 168°C (12 mmHg) | |
MFCD00009827 | |
1,4-bis 2-hydroxy-2-propyl benzene, 1,4-bis 1-methyl-1-hydroxyethyl benzene, alpha,alpha,alpha',alpha'-tetramethyl-1,4-benzenedimethanol, unii-g785drb5xh, p-bis 2-hydroxyisopropyl benzene, 1,4-bis 2-hydroxyisopropyl benzene, p-bis alpha-hydroxyisopropyl benzene, g785drb5xh, alpha,alpha'-dihydroxy-p-diisopropylbenzene, alpha,alpha,alpha',alpha'-tetramethyl-p-xylylenediol | |
LEARFTRDZQQTDN-UHFFFAOYSA-N | |
2-[4-(2-hydroxypropan-2-yl)phenyl]propan-2-ol | |
18062 | |
97% |
142°C to 146°C | |
C12H18O2 | |
1 g | |
Slightly Soluble in water (2.1g/L) (25°C). | |
CC(C)(C1=CC=C(C=C1)C(C)(C)O)O | |
194.274 | |
194.28 | |
1,4-Bis(2-hydroxyisopropyl)benzene |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 220-964-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only