missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,4-Bis(3-aminopropyl)piperazine 98.0+%, TCI America™
$61.38 - $318.52
Chemical Identifiers
CAS | 7209-38-3 |
---|---|
Molecular Formula | C10H24N4 |
Molecular Weight (g/mol) | 200.33 |
MDL Number | MFCD00006161 |
InChI Key | XUSNPFGLKGCWGN-UHFFFAOYSA-N |
Synonym | 1,4-bis 3-aminopropyl piperazine, 1,4-piperazinedipropanamine, 3,3'-piperazine-1,4-diyl bis propan-1-amine, bis aminopropyl piperazine, n,n'-bis 3-aminopropyl piperazine, unii-7gvx6c734b, bapp, ccris 8915, 1,4-di 3-aminopropyl piperazine, 3,3'-piperazine-1,4-diyldipropan-1-amine |
PubChem CID | 81629 |
IUPAC Name | 3-[4-(3-aminopropyl)piperazin-1-yl]propan-1-amine |
SMILES | C1CN(CCN1CCCN)CCCN |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
B104125ML
|
TCI America
B104125ML |
25 mL |
Each for $61.38
|
|
|||||
B1041500ML
|
TCI America
B1041500ML |
500 mL |
Each for $318.52
|
|
|||||
Chemical Identifiers
7209-38-3 | |
200.33 | |
XUSNPFGLKGCWGN-UHFFFAOYSA-N | |
81629 | |
C1CN(CCN1CCCN)CCCN |
C10H24N4 | |
MFCD00006161 | |
1,4-bis 3-aminopropyl piperazine, 1,4-piperazinedipropanamine, 3,3'-piperazine-1,4-diyl bis propan-1-amine, bis aminopropyl piperazine, n,n'-bis 3-aminopropyl piperazine, unii-7gvx6c734b, bapp, ccris 8915, 1,4-di 3-aminopropyl piperazine, 3,3'-piperazine-1,4-diyldipropan-1-amine | |
3-[4-(3-aminopropyl)piperazin-1-yl]propan-1-amine |
Specifications
7209-38-3 | |
158°C | |
MFCD00006161 | |
3267 | |
XUSNPFGLKGCWGN-UHFFFAOYSA-N | |
3-[4-(3-aminopropyl)piperazin-1-yl]propan-1-amine | |
81629 | |
≥98.0% (GC,T) | |
1,4-Bis(3-aminopropyl)piperazine |
Yellow | |
C10H24N4 | |
25 mL | |
1,4-bis 3-aminopropyl piperazine, 1,4-piperazinedipropanamine, 3,3'-piperazine-1,4-diyl bis propan-1-amine, bis aminopropyl piperazine, n,n'-bis 3-aminopropyl piperazine, unii-7gvx6c734b, bapp, ccris 8915, 1,4-di 3-aminopropyl piperazine, 3,3'-piperazine-1,4-diyldipropan-1-amine | |
C1CN(CCN1CCCN)CCCN | |
200.33 | |
200.33 | |
Liquid |
Safety and Handling
EINECSNumber : (5)-0962
TSCA : Yes