Learn More
1-[4-(Bromomethyl)phenyl]-1H-1,2,4-triazole, 97%, Thermo Scientific™
$235.91 - $551.04
Chemical Identifiers
CAS | 58419-69-5 |
---|---|
Molecular Formula | C9H8BrN3 |
Molecular Weight (g/mol) | 238.088 |
MDL Number | MFCD04113600 |
InChI Key | NOUAEJFAVOHNSY-UHFFFAOYSA-N |
Synonym | 1-4-bromomethyl phenyl-1h-1,2,4-triazole, 1-4-bromomethyl phenyl-1,2,4-triazole, 1h-1,2,4-triazole, 1-4-bromomethyl phenyl, 1-4-bromomethylphenyl-1h-1,2,4-triazole, zlchem 1040, 1-4-bromomethyl phenyl-1h-1,2,4-triazol, 1-4-bromomethyl-phenyl-1h-1,2,4-triazole, 1-4-bromo-methyl phenyl-1h-1,2,4-triazole |
PubChem CID | 2776489 |
IUPAC Name | 1-[4-(bromomethyl)phenyl]-1,2,4-triazole |
SMILES | C1=CC(=CC=C1CBr)N2C=NC=N2 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
CC18608CB
|
Thermo Scientific Chemicals
CC18608CB |
250 mg | Amber Glass Bottle |
Each for $235.91
|
|
|||||
CC18608DA
|
Thermo Scientific Chemicals
CC18608DA |
1 g | Amber Glass Bottle |
Each for $551.04
|
|
|||||
Chemical Identifiers
58419-69-5 | |
238.088 | |
NOUAEJFAVOHNSY-UHFFFAOYSA-N | |
2776489 | |
C1=CC(=CC=C1CBr)N2C=NC=N2 |
C9H8BrN3 | |
MFCD04113600 | |
1-4-bromomethyl phenyl-1h-1,2,4-triazole, 1-4-bromomethyl phenyl-1,2,4-triazole, 1h-1,2,4-triazole, 1-4-bromomethyl phenyl, 1-4-bromomethylphenyl-1h-1,2,4-triazole, zlchem 1040, 1-4-bromomethyl phenyl-1h-1,2,4-triazol, 1-4-bromomethyl-phenyl-1h-1,2,4-triazole, 1-4-bromo-methyl phenyl-1h-1,2,4-triazole | |
1-[4-(bromomethyl)phenyl]-1,2,4-triazole |
Specifications
58419-69-5 | |
100.0 | |
Amber Glass Bottle | |
MFCD04113600 | |
1-4-bromomethyl phenyl-1h-1,2,4-triazole, 1-4-bromomethyl phenyl-1,2,4-triazole, 1h-1,2,4-triazole, 1-4-bromomethyl phenyl, 1-4-bromomethylphenyl-1h-1,2,4-triazole, zlchem 1040, 1-4-bromomethyl phenyl-1h-1,2,4-triazol, 1-4-bromomethyl-phenyl-1h-1,2,4-triazole, 1-4-bromo-methyl phenyl-1h-1,2,4-triazole | |
C1=CC(=CC=C1CBr)N2C=NC=N2 | |
238.088 | |
319 | |
1-[4-(Bromomethyl)phenyl]-1H-1, 2, 4-triazole hydrobromide hydrate |
96.5 | |
97% | |
C9H8BrN3 | |
250 mg | |
NOUAEJFAVOHNSY-UHFFFAOYSA-N | |
1-[4-(bromomethyl)phenyl]-1,2,4-triazole | |
2776489 | |
97% |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger