missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-(4-Bromophenyl)hexane, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals BTB09866DA
Specifications
1-(4-Bromophenyl)hexane | |
96.5 | |
97% | |
C12H17Br | |
MFCD00061114 | |
1-4-bromophenyl hexane, 4-hexylbromobenzene, 1-bromo-4-n-hexylbenzene, 4-bromo-1-hexylbenzene, benzene, 1-bromo-4-hexyl, 4-bromo-n-hexylbenzene, 4-hexylphenyl bromide, 1-bromo-4-hexyl-benzene, acmc-1cb1g, # | |
CCCCCCC1=CC=C(Br)C=C1 | |
241.17 | |
241.17 |
23703-22-2 | |
100.0 | |
Amber Glass Bottle | |
BrC6H4(CH2)5CH3 | |
1 g | |
MXHOLIARBWJKCR-UHFFFAOYSA-N | |
1-bromo-4-hexylbenzene | |
598186 | |
97% |
Chemical Identifiers
23703-22-2 | |
241.17 | |
MXHOLIARBWJKCR-UHFFFAOYSA-N | |
598186 | |
CCCCCCC1=CC=C(Br)C=C1 |
C12H17Br | |
MFCD00061114 | |
1-4-bromophenyl hexane, 4-hexylbromobenzene, 1-bromo-4-n-hexylbenzene, 4-bromo-1-hexylbenzene, benzene, 1-bromo-4-hexyl, 4-bromo-n-hexylbenzene, 4-hexylphenyl bromide, 1-bromo-4-hexyl-benzene, acmc-1cb1g, # | |
1-bromo-4-hexylbenzene |