missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(4-Bromophenyl)octane, 97%, Thermo Scientific™

Chemical Identifiers

CAS 51554-93-9
Molecular Formula C14H21Br
Molecular Weight (g/mol) 269.226
MDL Number MFCD00038348
InChI Key OOZQSVXPBCINJF-UHFFFAOYSA-N
Synonym 1-4-bromophenyl octane, 1-bromo-4-n-octylbenzene, p-octylbromobenzene, p-bromophenyloctane, 4-n-octylbromobenzene, 1-p-bromophenyl octane, 4-bromooctylbenzene, 4-oct-1-yl bromobenzene, benzene, 1-bromo-4-octyl, 4-bromo-1-octylbenzene
PubChem CID 142854
IUPAC Name 1-bromo-4-octylbenzene
SMILES CCCCCCCCC1=CC=C(C=C1)Br
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
BTB09864DA
SDS View Documents
Thermo Scientific Chemicals
BTB09864DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BTB09864EA
SDS View Documents
Thermo Scientific Chemicals
BTB09864EA
10g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BTB09864EE
SDS View Documents
Thermo Scientific Chemicals
BTB09864EE
50g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

51554-93-9
269.226
OOZQSVXPBCINJF-UHFFFAOYSA-N
142854
CCCCCCCCC1=CC=C(C=C1)Br
C14H21Br
MFCD00038348
1-4-bromophenyl octane, 1-bromo-4-n-octylbenzene, p-octylbromobenzene, p-bromophenyloctane, 4-n-octylbromobenzene, 1-p-bromophenyl octane, 4-bromooctylbenzene, 4-oct-1-yl bromobenzene, benzene, 1-bromo-4-octyl, 4-bromo-1-octylbenzene
1-bromo-4-octylbenzene

Specifications

51554-93-9
100.0
Amber Glass Bottle
BrC6H4(CH2)7CH3
1g
OOZQSVXPBCINJF-UHFFFAOYSA-N
1-bromo-4-octylbenzene
142854
97%
96.5
97%
C14H21Br
MFCD00038348
1-4-bromophenyl octane, 1-bromo-4-n-octylbenzene, p-octylbromobenzene, p-bromophenyloctane, 4-n-octylbromobenzene, 1-p-bromophenyl octane, 4-bromooctylbenzene, 4-oct-1-yl bromobenzene, benzene, 1-bromo-4-octyl, 4-bromo-1-octylbenzene
CCCCCCCCC1=CC=C(C=C1)Br
269.226
269.22
1-(4-Bromophenyl)octane
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'