missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(4-chlorophenyl)-5-methyl-1H-pyrazole-4-carbonyl chloride, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC00081CB

 View more versions of this product

Catalog No. CC00081CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

1-(4-chlorophenyl)-5-methyl-1H-pyrazole-4-carbonyl chloride
96.5
97%
C11H8Cl2N2O
250mg
KUQHQFYKJPXTNB-UHFFFAOYSA-N
1-(4-chlorophenyl)-5-methylpyrazole-4-carbonyl chloride
2776128
97%
288252-38-0
100.0
Amber glass bottle
MFCD03407325
1-4-chlorophenyl-5-methyl-1h-pyrazole-4-carbonyl chloride, 1-4-chlorophenyl-5-methylpyrazole-4-carbonyl chloride, 1-4-chlorophenyl-5-methyl-1h-pyrazole-4-carbonylchloride, 1-4-chlorophenyl-5-methylpyrazole-4-carboxylic chloride, 1-4-chloro-phenyl-5-methyl-1h-pyrazole-4-carbonyl chloride, 1h-pyrazole-4-carbonylchloride, 1-4-chlorophenyl-5-methyl
CC1=C(C=NN1C2=CC=C(C=C2)Cl)C(=O)Cl
255.098
255.1

Chemical Identifiers

288252-38-0
255.098
KUQHQFYKJPXTNB-UHFFFAOYSA-N
2776128
CC1=C(C=NN1C2=CC=C(C=C2)Cl)C(=O)Cl
C11H8Cl2N2O
MFCD03407325
1-4-chlorophenyl-5-methyl-1h-pyrazole-4-carbonyl chloride, 1-4-chlorophenyl-5-methylpyrazole-4-carbonyl chloride, 1-4-chlorophenyl-5-methyl-1h-pyrazole-4-carbonylchloride, 1-4-chlorophenyl-5-methylpyrazole-4-carboxylic chloride, 1-4-chloro-phenyl-5-methyl-1h-pyrazole-4-carbonyl chloride, 1h-pyrazole-4-carbonylchloride, 1-4-chlorophenyl-5-methyl
1-(4-chlorophenyl)-5-methylpyrazole-4-carbonyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions