missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Cyclohexanediol (cis- and trans- mixture) 99.0+%, TCI America™
$37.29 - $326.21
Chemical Identifiers
| CAS | 556-48-9 |
|---|---|
| Molecular Formula | C6H12O2 |
| Molecular Weight (g/mol) | 116.16 |
| MDL Number | MFCD00001448,MFCD00063612,MFCD00075462 |
| InChI Key | VKONPUDBRVKQLM-UHFFFAOYSA-N |
| Synonym | 1,4-cyclohexanediol, cis-1,4-cyclohexanediol, trans-1,4-cyclohexanediol, trans-cyclohexane-1,4-diol, quinitol, 1,4-cyclohexanediol, trans, cis-cyclohexane-1,4-diol, 1,4-cyclohexanediol, cis, trans-1,4-dihydroxycyclohexane, cis-1,4-dihydroxycyclohexane |
| PubChem CID | 11162 |
| IUPAC Name | cyclohexane-1,4-diol |
| SMILES | OC1CCC(O)CC1 |
Chemical Identifiers
| 556-48-9 | |
| 116.16 | |
| VKONPUDBRVKQLM-UHFFFAOYSA-N | |
| 11162 | |
| OC1CCC(O)CC1 |
| C6H12O2 | |
| MFCD00001448,MFCD00063612,MFCD00075462 | |
| 1,4-cyclohexanediol, cis-1,4-cyclohexanediol, trans-1,4-cyclohexanediol, trans-cyclohexane-1,4-diol, quinitol, 1,4-cyclohexanediol, trans, cis-cyclohexane-1,4-diol, 1,4-cyclohexanediol, cis, trans-1,4-dihydroxycyclohexane, cis-1,4-dihydroxycyclohexane | |
| cyclohexane-1,4-diol |
Specifications
| 556-48-9 | |
| White | |
| C6H12O2 | |
| 25 g | |
| VKONPUDBRVKQLM-UHFFFAOYSA-N | |
| cyclohexane-1,4-diol | |
| 11162 | |
| ≥99.0% (GC) | |
| 1,4-Cyclohexanediol (cis- and trans- mixture) |
| 101°C | |
| 132°C | |
| MFCD00001448,MFCD00063612,MFCD00075462 | |
| 1,4-cyclohexanediol, cis-1,4-cyclohexanediol, trans-1,4-cyclohexanediol, trans-cyclohexane-1,4-diol, quinitol, 1,4-cyclohexanediol, trans, cis-cyclohexane-1,4-diol, 1,4-cyclohexanediol, cis, trans-1,4-dihydroxycyclohexane, cis-1,4-dihydroxycyclohexane | |
| OC1CCC(O)CC1 | |
| 116.16 | |
| 116.16 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-2310
TSCA : No