Learn More
1,4-Diazabicyclo[2.2.2]octane, 98%, Thermo Scientific Chemicals
$47.08 - $321.63
Chemical Identifiers
CAS | 280-57-9 |
---|---|
Molecular Formula | C6H12N2 |
Molecular Weight (g/mol) | 112.176 |
MDL Number | MFCD00006689 |
InChI Key | IMNIMPAHZVJRPE-UHFFFAOYSA-N |
Synonym | 1,4-diazabicyclo 2.2.2 octane, triethylenediamine, dabco, dabco 33lv, 1,4-ethylenepiperazine, dabco crystal, teda, texacat td 100, dabco s-25 |
PubChem CID | 9237 |
IUPAC Name | 1,4-diazabicyclo[2.2.2]octane |
SMILES | C1CN2CCN1CC2 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1400314
|
Thermo Scientific Chemicals
A1400314 |
25 g |
Each for $47.08
|
|
|||||
AAA1400322
|
Thermo Scientific Chemicals
A1400322 |
100 g |
Each for $116.49
|
|
|||||
AAA1400336
|
Thermo Scientific Chemicals
A1400336 |
500 g |
Each for $321.63
|
|
|||||
Description
1,4-Diazabicyclo[2.2.2]octane is used as polyurethane catalyst, Balis-Hillman reaction catalyst complexing ligand and lewis base. It finds use in dye lasers and in mounting samples for fluorescence microscopy and as anti-fade reagent shown to scavenge free radicals due to flurochrome excitation of fluorochromes. Further, it is an oxidation and polymerization catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,4-Diazabicyclo[2.2.2]octane is used as polyurethane catalyst, Balis-Hillman reaction catalyst complexing ligand and lewis base. It finds use in dye lasers and in mounting samples for fluorescence microscopy and as anti-fade reagent shown to scavenge free radicals due to flurochrome excitation of fluorochromes. Further, it is an oxidation and polymerization catalyst.
Solubility
Soluble in water, acetone, benzene, ethanol and methyl ethyl ketone.
Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids.
Chemical Identifiers
280-57-9 | |
112.176 | |
IMNIMPAHZVJRPE-UHFFFAOYSA-N | |
9237 | |
C1CN2CCN1CC2 |
C6H12N2 | |
MFCD00006689 | |
1,4-diazabicyclo 2.2.2 octane, triethylenediamine, dabco, dabco 33lv, 1,4-ethylenepiperazine, dabco crystal, teda, texacat td 100, dabco s-25 | |
1,4-diazabicyclo[2.2.2]octane |
Specifications
280-57-9 | |
1.14 | |
62°C (143°F) | |
C6H12N2 | |
25 g | |
103618 | |
14,9669 | |
Soluble in water,acetone,benzene,ethanol and methyl ethyl ketone. | |
C1CN2CCN1CC2 | |
112.176 | |
112.17 | |
1,4-Diazabicyclo[2.2.2]octane |
155°C to 160°C | |
174°C to 176°C | |
Amine-like | |
MFCD00006689 | |
UN1325 | |
Hygroscopic | |
1,4-diazabicyclo 2.2.2 octane, triethylenediamine, dabco, dabco 33lv, 1,4-ethylenepiperazine, dabco crystal, teda, texacat td 100, dabco s-25 | |
IMNIMPAHZVJRPE-UHFFFAOYSA-N | |
1,4-diazabicyclo[2.2.2]octane | |
9237 | |
98% |
Safety and Handling
GHS H Statement
H228-H318-H302-H335-H315
Flammable solid.
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
P210-P240-P241-P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P310-P330-P332+P313-P362-P370+P378q-P501c
H228-H302-H315-H318-H500
DOTInformation : Hazard Class: 4.1; Packaging Group: II
EINECSNumber : 205-999-9
RTECSNumber : HM0354200
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only