Learn More
1,4-Dibromo-2,5-difluorobenzene, 98%
CAS: 327-51-5 | C6H2Br2F2 | 271.887 g/mol
$74.51 - $970.92
Chemical Identifiers
| CAS | 327-51-5 |
|---|---|
| Molecular Formula | C6H2Br2F2 |
| Molecular Weight (g/mol) | 271.887 |
| MDL Number | MFCD00000346 |
| InChI Key | GLVMLJCMUBZVTJ-UHFFFAOYSA-N |
| Synonym | 1,4-dibromo-2,5-difluoro-benzene, benzene, 1,4-dibromo-2,5-difluoro, 2,5-difluoro-1,4-dibromo benzene, maybridge1_003946, pubchem3441, acmc-1cmi9, chemwish ic06783, ksc495q1l, emolecules 493137, 2,5-dibromo-1,4-difluorobenzene |
| PubChem CID | 67596 |
| IUPAC Name | 1,4-dibromo-2,5-difluorobenzene |
| SMILES | C1=C(C(=CC(=C1Br)F)Br)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1424806
|
Thermo Scientific Chemicals
A1424806 |
5 g |
Each for $74.51
|
|
|||||
|
AAA1424814
|
Thermo Scientific Chemicals
A1424814 |
25 g |
Each for $247.14
|
|
|||||
|
AAA1424822
|
Thermo Scientific Chemicals
A1424822 |
100 g |
Each for $970.92
|
|
|||||
Description
1,4-Dibromo-2,5-difluorobenzene has been used in the preparation of 1,2,4,5-tetrakis(phosphino)-3,6-difluorobenzenes and poly(2,5-difluoro-2,5-didodecyl-4,4-biphenylylene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,4-Dibromo-2,5-difluorobenzene has been used in the preparation of 1,2,4,5-tetrakis(phosphino)-3,6-difluorobenzenes and poly(2,5-difluoro-2′,5′-didodecyl-4,4′-biphenylylene.
Solubility
Soluble in methanol. Insoluble in water.
Notes
Stable under recommended storage conditions. Store in cool, dry place in tightly closed containers. With good ventilation. Store away from oxidizing agent.
Chemical Identifiers
| 327-51-5 | |
| 271.887 | |
| GLVMLJCMUBZVTJ-UHFFFAOYSA-N | |
| 67596 | |
| C1=C(C(=CC(=C1Br)F)Br)F |
| C6H2Br2F2 | |
| MFCD00000346 | |
| 1,4-dibromo-2,5-difluoro-benzene, benzene, 1,4-dibromo-2,5-difluoro, 2,5-difluoro-1,4-dibromo benzene, maybridge1_003946, pubchem3441, acmc-1cmi9, chemwish ic06783, ksc495q1l, emolecules 493137, 2,5-dibromo-1,4-difluorobenzene | |
| 1,4-dibromo-2,5-difluorobenzene |
Specifications
| 327-51-5 | |
| 96°C (20 mmHg) | |
| MFCD00000346 | |
| 2250095 | |
| Soluble in methanol. Insoluble in water. | |
| C1=C(C(=CC(=C1Br)F)Br)F | |
| 271.887 | |
| 271.89 | |
| 1,4-Dibromo-2,5-difluorobenzene |
| 60°C to 64°C | |
| C6H2Br2F2 | |
| 5 g | |
| 1,4-dibromo-2,5-difluoro-benzene, benzene, 1,4-dibromo-2,5-difluoro, 2,5-difluoro-1,4-dibromo benzene, maybridge1_003946, pubchem3441, acmc-1cmi9, chemwish ic06783, ksc495q1l, emolecules 493137, 2,5-dibromo-1,4-difluorobenzene | |
| GLVMLJCMUBZVTJ-UHFFFAOYSA-N | |
| 1,4-dibromo-2,5-difluorobenzene | |
| 67596 | |
| 98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 206-317-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only