Learn More
1,4-Dibromobenzene, 99%
CAS: 106-37-6 | C6H4Br2 | 235.91 g/mol
$157.34 - $485.53
Chemical Identifiers
| CAS | 106-37-6 |
|---|---|
| Molecular Formula | C6H4Br2 |
| Molecular Weight (g/mol) | 235.91 |
| MDL Number | MFCD00000089 |
| InChI Key | SWJPEBQEEAHIGZ-UHFFFAOYSA-N |
| Synonym | p-dibromobenzene, p-bromophenyl bromide, benzene, 1,4-dibromo, benzene, p-dibromo, p-benzene dibromide, 1,4-dibromo benzene, 1,4-dibromo-benzene, unii-9991w3m5hz, dibromobenzene, 1,4, labotest-bb ltbb002071 |
| PubChem CID | 7804 |
| ChEBI | CHEBI:37150 |
| IUPAC Name | 1,4-dibromobenzene |
| SMILES | BrC1=CC=C(Br)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC112732500
|
Thermo Scientific Chemicals
112732500 |
250 g | Plastic bottle |
Each for $157.34
|
|
||||
|
AC112730010
|
Thermo Scientific Chemicals
112730010 |
1 kg | Plastic bottle |
Each for $485.53
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 106-37-6 | |
| 235.91 | |
| SWJPEBQEEAHIGZ-UHFFFAOYSA-N | |
| 7804 | |
| 1,4-dibromobenzene |
| C6H4Br2 | |
| MFCD00000089 | |
| p-dibromobenzene, p-bromophenyl bromide, benzene, 1,4-dibromo, benzene, p-dibromo, p-benzene dibromide, 1,4-dibromo benzene, 1,4-dibromo-benzene, unii-9991w3m5hz, dibromobenzene, 1,4, labotest-bb ltbb002071 | |
| CHEBI:37150 | |
| BrC1=CC=C(Br)C=C1 |
Specifications
| 106-37-6 | |
| 100.0 | |
| Colorless to White | |
| 219°C | |
| Authentic | |
| Plastic bottle | |
| C6H4Br2 | |
| 250 g | |
| 1.84 | |
| p-dibromobenzene, p-bromophenyl bromide, benzene, 1,4-dibromo, benzene, p-dibromo, p-benzene dibromide, 1,4-dibromo benzene, 1,4-dibromo-benzene, unii-9991w3m5hz, dibromobenzene, 1,4, labotest-bb ltbb002071 | |
| SWJPEBQEEAHIGZ-UHFFFAOYSA-N | |
| 1,4-dibromobenzene | |
| 7804 | |
| 235.91 | |
| Crystals |
| 98.5 | |
| 86°C to 89°C | |
| 1.8400g/mL | |
| 100°C | |
| 98.5% min. (GC) | |
| C6H4Br2 | |
| MFCD00000089 | |
| 05, 211 | |
| 15, 3024 | |
| Solubility in water: practically insoluble. Other solubilities: soluble in benzene,chloroform and ether | |
| BrC1=CC=C(Br)C=C1 | |
| 235.91 | |
| CHEBI:37150 | |
| 99% | |
| 1, 4-Dibromobenzene |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO – Research Use Only