Learn More
1,4-Dichlorobutane, 99%, Thermo Scientific Chemicals
$8.39 - $256.85
Chemical Identifiers
CAS | 110-56-5 |
---|---|
Molecular Formula | C4H8Cl2 |
Molecular Weight (g/mol) | 127.01 |
MDL Number | MFCD00001011 |
InChI Key | KJDRSWPQXHESDQ-UHFFFAOYSA-N |
Synonym | butane, 1,4-dichloro, unii-8326ynm2b3, tetramethylene chloride, 1,4-dichlorobutane, butane,4-dichloro, tetramethylene dichloride, 1,4-bis chloranyl butane, ksc491k8b, 1,4-dichlorobutane, puriss gc |
PubChem CID | 8059 |
IUPAC Name | 1,4-dichlorobutane |
SMILES | ClCCCCCl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC216431000
|
Thermo Scientific Chemicals
216431000 |
100 mL | Glass bottle |
Each for $8.39
|
|
||||
AC216430010
|
Thermo Scientific Chemicals
216430010 |
1 L | Glass bottle |
Each for $256.85
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
110-56-5 | |
127.01 | |
KJDRSWPQXHESDQ-UHFFFAOYSA-N | |
8059 | |
ClCCCCCl |
C4H8Cl2 | |
MFCD00001011 | |
butane, 1,4-dichloro, unii-8326ynm2b3, tetramethylene chloride, 1,4-dichlorobutane, butane,4-dichloro, tetramethylene dichloride, 1,4-bis chloranyl butane, ksc491k8b, 1,4-dichlorobutane, puriss gc | |
1,4-dichlorobutane |
Specifications
110-56-5 | |
100.0 | |
1.1600g/mL | |
40°C | |
98.5% min. (GC) | |
C4H8Cl2 | |
Cl(CH2)4Cl | |
100 mL | |
1.16 | |
Solubility in water: 0.24% (20 c). Other solubilities: soluble in most common organic solvents | |
ClCCCCCl | |
127.01 | |
1.46 mPa.s (20°C) | |
99% | |
1, 4-Dichlorobutane, 99+% |
98.5 | |
-38°C | |
161°C to 163°C | |
Authentic | |
Glass bottle | |
1.4530 to 1.455 | |
MFCD00001011 | |
01, 119 | |
butane, 1,4-dichloro, unii-8326ynm2b3, tetramethylene chloride, 1,4-dichlorobutane, butane,4-dichloro, tetramethylene dichloride, 1,4-bis chloranyl butane, ksc491k8b, 1,4-dichlorobutane, puriss gc | |
KJDRSWPQXHESDQ-UHFFFAOYSA-N | |
1,4-dichlorobutane | |
8059 | |
127.01 | |
Liquid |
Safety and Handling
GHS H Statement:
Harmful to aquatic life with long lasting effects.
Flammable liquid and vapor.
GHS P Statement:
Avoid release to the environment.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 203-778-1
TSCA : TSCA
RUO – Research Use Only