missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,4-Dicyanobenzene, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 113820250

 View more versions of this product

Catalog No. AC113820250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1, 4-Dicyanobenzene
97.5
222°C to 227°C
Authentic
Glass bottle
C6H4(CN)2
25 g
terephthalonitrile, 1,4-dicyanobenzene, 1,4-benzenedicarbonitrile, p-phthalodinitrile, 4-cyanobenzonitrile, p-dicyanobenzene, p-benzenedinitrile, p-pdn, terephthalodinitrile, 1,4-benzodinitrile
BHXFKXOIODIUJO-UHFFFAOYSA-N
benzene-1,4-dicarbonitrile
12172
98%
623-26-7
100.0
White to Yellow
97.5% min. (GC)
C8H4N2
MFCD00001810
09, 846
Solubility in water: 0.08g/L (23°C). Other solubilities: soluble 15.3g/L in acetone (20°C),soluble in methanol
C1=CC(=CC=C1C#N)C#N
128.13
128.13
Powder

Chemical Identifiers

623-26-7
128.13
BHXFKXOIODIUJO-UHFFFAOYSA-N
12172
C1=CC(=CC=C1C#N)C#N
C8H4N2
MFCD00001810
terephthalonitrile, 1,4-dicyanobenzene, 1,4-benzenedicarbonitrile, p-phthalodinitrile, 4-cyanobenzonitrile, p-dicyanobenzene, p-benzenedinitrile, p-pdn, terephthalodinitrile, 1,4-benzodinitrile
benzene-1,4-dicarbonitrile
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 210-783-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only