missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,4-Diethylbenzene, 98%, Thermo Scientific Chemicals
$105.86 - $105.86
Chemical Identifiers
CAS | 105-05-5 |
---|---|
Molecular Formula | C10H14 |
Molecular Weight (g/mol) | 134.22 |
MDL Number | MFCD00009264 |
InChI Key | DSNHSQKRULAAEI-UHFFFAOYSA-N |
Synonym | p-diethylbenzene, benzene, 1,4-diethyl, p-ethylethylbenzene, benzene, p-diethyl, para-diethylbenzene, unii-0psm16x42d, acmc-1bu3z, dsstox_cid_6711, dsstox_rid_78192, dsstox_gsid_26711 |
PubChem CID | 7734 |
ChEBI | CHEBI:34062 |
IUPAC Name | 1,4-diethylbenzene |
SMILES | CCC1=CC=C(CC)C=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC114150050
|
Thermo Scientific Chemicals
114150050 |
5 mL | Glass bottle |
Each for $105.86
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
105-05-5 | |
134.22 | |
DSNHSQKRULAAEI-UHFFFAOYSA-N | |
7734 | |
1,4-diethylbenzene |
C10H14 | |
MFCD00009264 | |
p-diethylbenzene, benzene, 1,4-diethyl, p-ethylethylbenzene, benzene, p-diethyl, para-diethylbenzene, unii-0psm16x42d, acmc-1bu3z, dsstox_cid_6711, dsstox_rid_78192, dsstox_gsid_26711 | |
CHEBI:34062 | |
CCC1=CC=C(CC)C=C1 |
Specifications
105-05-5 | |
100.0 | |
Colorless | |
184°C | |
Authentic | |
Glass bottle | |
1.4935 to 1.4955 | |
MFCD00009264 | |
05, 426 | |
p-diethylbenzene, benzene, 1,4-diethyl, p-ethylethylbenzene, benzene, p-diethyl, para-diethylbenzene, unii-0psm16x42d, acmc-1bu3z, dsstox_cid_6711, dsstox_rid_78192, dsstox_gsid_26711 | |
DSNHSQKRULAAEI-UHFFFAOYSA-N | |
1,4-diethylbenzene | |
7734 | |
134.22 | |
Liquid |
97.5 | |
-43°C | |
0.8600g/mL | |
56°C | |
97.5% min. (GC) | |
C10H14 | |
C6H4(C2H5)2 | |
5 mL | |
0.86 | |
Solubility in water: insoluble in water. Other solubilities: soluble in benzene and carbon tetrachloride,miscible with ethanol,ether and acetone | |
CCC1=CC=C(CC)C=C1 | |
134.22 | |
CHEBI:34062 | |
98% | |
1, 4-Diethylbenzene, 98% |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 203-265-2
TSCA : TSCA
RUO – Research Use Only