missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,4-Diethylbenzene, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 114150250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
1, 4-Diethylbenzene | |
105-05-5 | |
100.0 | |
184°C | |
Authentic | |
Glass bottle | |
1.4935 to 1.4955 | |
MFCD00009264 | |
05, 426 | |
p-diethylbenzene, benzene, 1,4-diethyl, p-ethylethylbenzene, benzene, p-diethyl, para-diethylbenzene, unii-0psm16x42d, acmc-1bu3z, dsstox_cid_6711, dsstox_rid_78192, dsstox_gsid_26711 | |
DSNHSQKRULAAEI-UHFFFAOYSA-N | |
1,4-diethylbenzene | |
7734 | |
134.22 |
98% | |
97.5 | |
0.8600g/mL | |
56°C | |
97.5% min. (GC) | |
C10H14 | |
C6H4(C2H5)2 | |
25mL | |
0.86 | |
Solubility in water: insoluble in water. Other solubilities: soluble in benzene and carbon tetrachloride,miscible with ethanol,ether and acetone | |
CCC1=CC=C(CC)C=C1 | |
134.22 | |
CHEBI:34062 | |
98% |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 203-265-2
TSCA : TSCA
RUO â Research Use Only