missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,4-Difluorobenzene, 99+%, Thermo Scientific Chemicals

$154.64 - $154.64

Chemical Identifiers

CAS 540-36-3
Molecular Formula C6H4F2
Molecular Weight (g/mol) 114.10
MDL Number MFCD00000344
InChI Key QUGUFLJIAFISSW-UHFFFAOYSA-N
Synonym p-difluorobenzene, benzene, 1,4-difluoro, para-difluorobenzene, benzene, p-difluoro, 2,5-difluorobenzene, unii-y9fq2ef0z6, 1,4-difluorbenzol, y9fq2ef0z6, benzene,4-difluoro, pubchem2309
PubChem CID 10892
ChEBI CHEBI:38585
IUPAC Name 1,4-difluorobenzene
SMILES FC1=CC=C(F)C=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC114610250
SDS View Documents
Thermo Scientific Chemicals
114610250
25 mL Glass bottle
Each for $154.64
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

540-36-3
114.10
QUGUFLJIAFISSW-UHFFFAOYSA-N
10892
1,4-difluorobenzene
C6H4F2
MFCD00000344
p-difluorobenzene, benzene, 1,4-difluoro, para-difluorobenzene, benzene, p-difluoro, 2,5-difluorobenzene, unii-y9fq2ef0z6, 1,4-difluorbenzol, y9fq2ef0z6, benzene,4-difluoro, pubchem2309
CHEBI:38585
FC1=CC=C(F)C=C1

Specifications

540-36-3
100.0
Colorless to Yellow
88°C to 89°C
Authentic
Glass bottle
1.4400 to 1.442
MFCD00000344
05, 199
15, 3167
QUGUFLJIAFISSW-UHFFFAOYSA-N
1,4-difluorobenzene
10892
114.09
Liquid
99.0
-13°C
1.1700g/mL
2°C
99+%
C6H4F2
C6H4F2
25 mL
1.17
p-difluorobenzene, benzene, 1,4-difluoro, para-difluorobenzene, benzene, p-difluoro, 2,5-difluorobenzene, unii-y9fq2ef0z6, 1,4-difluorbenzol, y9fq2ef0z6, benzene,4-difluoro, pubchem2309
FC1=CC=C(F)C=C1
114.10
CHEBI:38585
99+%
1, 4-Difluorobenzene, 99+%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Highly flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 208-742-9

RTECSNumber : CZ5658000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only