missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,4-Dinitrobenzene, 98+%, Thermo Scientific Chemicals

$101.73 - $1329.45

Chemical Identifiers

CAS 100-25-4
Molecular Formula C6H4N2O4
Molecular Weight (g/mol) 168.108
MDL Number MFCD00007314
InChI Key FYFDQJRXFWGIBS-UHFFFAOYSA-N
Synonym p-dinitrobenzene, benzene, 1,4-dinitro, benzene, p-dinitro, dithane a-4, dinitrobenzene, p, dinitrobenzene, para, ccris 3092, 1,4-dinitro-benzene, 1,4-dinitrobenzene, analytical standard, para-dinitrobenzene
PubChem CID 7492
ChEBI CHEBI:51398
IUPAC Name 1,4-dinitrobenzene
SMILES C1=CC(=CC=C1[N+](=O)[O-])[N+](=O)[O-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1029306
View Documents
Thermo Scientific Chemicals
A1029306
5 g
Each for $101.73
 
AAA1029314
View Documents
Thermo Scientific Chemicals
A1029314
25 g
Each for $336.01
 
AAA1029322
View Documents
Thermo Scientific Chemicals
A1029322
100 g
Each for $1,329.45
 
Description

Description

1,4-Dinitrobenzene is used in a study to evaluate the ionization mechanism and solvent effect by novel atmospheric pressure photoionization mass spectrometry in negative ion mode for analysis of some compounds. 1,4-Dinitrobenzene can be used in synthesis of dyes and dye intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Dinitrobenzene is used in a study to evaluate the ionization mechanism and solvent effect by novel atmospheric pressure photoionization mass spectrometry in negative ion mode for analysis of some compounds. 1,4-Dinitrobenzene can be used in synthesis of dyes and dye intermediates.

Solubility
Soluble in water. (0.8 g/L) at 20°C.

Notes
Incompatible with bases, alkali metals, oxidizing agents and nitric acid.
Specifications

Chemical Identifiers

100-25-4
168.108
FYFDQJRXFWGIBS-UHFFFAOYSA-N
7492
1,4-dinitrobenzene
C6H4N2O4
MFCD00007314
p-dinitrobenzene, benzene, 1,4-dinitro, benzene, p-dinitro, dithane a-4, dinitrobenzene, p, dinitrobenzene, para, ccris 3092, 1,4-dinitro-benzene, 1,4-dinitrobenzene, analytical standard, para-dinitrobenzene
CHEBI:51398
C1=CC(=CC=C1[N+](=O)[O-])[N+](=O)[O-]

Specifications

100-25-4
1.625
150°C (302°F)
MFCD00007314
UN3443
14,3273
Soluble in water. (0.8g/L) at 20°C.
C1=CC(=CC=C1[N+](=O)[O-])[N+](=O)[O-]
168.108
CHEBI:51398
≥98%
172°C to 176°C
299°C
C6H4N2O4
5 g
1105828
p-dinitrobenzene, benzene, 1,4-dinitro, benzene, p-dinitro, dithane a-4, dinitrobenzene, p, dinitrobenzene, para, ccris 3092, 1,4-dinitro-benzene, 1,4-dinitrobenzene, analytical standard, para-dinitrobenzene
FYFDQJRXFWGIBS-UHFFFAOYSA-N
1,4-dinitrobenzene
7492
168.11
1,4-Dinitrobenzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.

P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P501c

H300+H310+H330-H373

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: DINITROBENZENES, SOLID

EINECSNumber : 202-833-7

RTECSNumber : CZ7525000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only