missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Dioxane (Certified ACS), Fisher Chemical™
$1852.27 - $1852.27
Chemical Identifiers
| CAS | 123-91-1 |
|---|---|
| Molecular Formula | C4H8O2 |
| Molecular Weight (g/mol) | 88.106 |
| MDL Number | MFCD00006571 |
| InChI Key | RYHBNJHYFVUHQT-UHFFFAOYSA-N |
| Synonym | dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin |
| PubChem CID | 31275 |
| ChEBI | CHEBI:47032 |
| IUPAC Name | 1,4-dioxane |
| SMILES | C1COCCO1 |
Chemical Identifiers
| 123-91-1 | |
| 88.106 | |
| RYHBNJHYFVUHQT-UHFFFAOYSA-N | |
| 31275 | |
| 1,4-dioxane |
| C4H8O2 | |
| MFCD00006571 | |
| dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin | |
| CHEBI:47032 | |
| C1COCCO1 |
Specifications
| 6 to 8 | |
| 123-91-1 | |
| 1.034g/cm³ | |
| 4 L | |
| Glass Bottle | |
| 0.004% max. | |
| 0.0016mEq/g max. | |
| MFCD00006571 | |
| 1.32 mPa/s at 20°C | |
| dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin | |
| C1COCCO1 | |
| 88.106 | |
| CHEBI:47032 | |
| ≥99% | |
| 1, 4-Dioxane |
| 12°C | |
| Colorless | |
| 101°C | |
| 12°C (53.6°F) | |
| Liquid | |
| ≥99 % | |
| C4H8O2 | |
| 41mbar at 20°C | |
| 0.02% max. | |
| RYHBNJHYFVUHQT-UHFFFAOYSA-N | |
| 1,4-dioxane | |
| 31275 | |
| 88.11 | |
| Certified ACS Reagent |
Safety and Handling
DANGER!
Emergency Overview
Flammable liquid and vapor. May form explosive peroxides. Possible cancer hazard. May cause cancer based on animal data. Irritating to eyes, respiratory system and skin. Inhalation may cause central nervous system effects. Repeated exposure may cause skin dryness or cracking. Hygroscopic.
NFPA
Health: 2
Flammability: 3
Instability: 1
SIGNAL WORD: DANGER!
DOTInformation : DOT Class 3, : Flammable Liquid